Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00612997_m1
          See other STBD1 GE Assays ›
          Gene Symbol:
          Stbd1
          Gene Name:
          starch binding domain 1
          Gene Aliases:
          D530019K15Rik, D5Ertd593e
          Chromosome Location:
          Chr.5: 92603051 - 92606579 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00612997_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stbd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175096.3 NP_780305.1 1 - 2 285 83
          GenBank mRNA AK050416.1 - 1 - 2 286 83
          BC052666.1 - 1 - 2 246 83
          BC118661.1 - 1 - 2 268 40107321 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500000877.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stbd1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          carbohydrate binding
          starch binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23895034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23895033 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23895036 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23895032 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967231 BC052666.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967231 NM_175096.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967231 AK050416.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470919 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112103 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967231 BC052666.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967231 NM_175096.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967231 AK050416.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895034 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895038 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895016 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23895022 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112103 - -
          Whole Mouse Genome G4122A A_52_P645855 BC052666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P645855 BC118615.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P645855 BC118661.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P645855 NM_175096.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P645855 AK050416.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119919 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112103 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04274885 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04274886 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23895033 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23895034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23895032 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23895036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23895011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23894992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895064 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04922492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04445845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04922524 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04658618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04922522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04286191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895055 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23894987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895067 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895030 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895024 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895033 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04835576 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04164896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04445841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04922506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04922502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04164897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04922490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04835580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04835582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04835583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23895034 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10523257 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523259 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523256 - -
          Affymetrix Mouse Gene 1.0 ST Array 10523258 - -
          Affymetrix HT MG-430 PM Array Plate 1434442_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10523255 - -
          Mouse Genome 430 2.0 Array 1434442_at - -
          Mouse Genome 430 2.0 Array 1434442_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52331_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 52331_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500007635.mm.1 TC0500000877.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007635.mm.1 NM_175096.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007635.mm.1 NM_175096.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007638.mm.1 TC0500000877.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007638.mm.1 NM_175096.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007638.mm.1 NM_175096.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007639.mm.1 TC0500000877.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500007639.mm.1 NM_175096.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500007639.mm.1 NM_175096.3 Refseq
          Mouse Expression Set 430 1434442_PM_at - -
          Mouse Expression Set 430 1434442_at - -
          Murine Genome U74v2 Set 104116_PM_at - -
          Murine Genome U74v2 Set 104116_at - -
          Mouse Genome 430A 2.0 Array 1434442_at - -
          Mouse Genome 430A 2.0 Array 1434442_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434442_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1434442_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 570706 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1400286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001400286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2663211 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2663211 - -
          mouse-6 v1.1 (Illumina) scl27580.3_53-S - -
          Illumina Sentrix mouse-6.v1 bead array 1400286 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27580.3_53-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1400286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2663211 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2663211 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl27580.3_53-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline