Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00613213_m1
          See other SRCAP GE Assays ›
          Gene Symbol:
          Srcap
          Gene Name:
          Snf2-related CREBBP activator protein
          Gene Aliases:
          B930091H02Rik, D030022P06Rik, F630004O05Rik
          Chromosome Location:
          Chr.7: 127511983 - 127561219 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00613213_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Srcap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK051146.1 - 4 - 5 905 75
          AK142089.1 - 4 - 5 949 75
          AK154145.1 - 5 - 6 590 75
          AK169954.1 - 4 - 5 933 75
          BC085144.1 - 4 - 5 888 75

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P419632 BC085144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P419632 AK142089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P419632 AK154145.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P419632 AK169954.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P419632 AK051146.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P419632 BC085144.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P419632 AK142089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P419632 AK154145.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P419632 AK169954.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P419632 AK051146.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002497 BC048731.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002497 AK081765.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2002497 AA881307.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126225 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146758 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109907 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407789 BC085144.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407789 AK142089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407789 AK169954.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407789 AK051146.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P407789 BC049754.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002497 BC048731.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002497 AK081765.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2002497 AA881307.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146758 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06361848 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109907 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126225 - -
          Whole Mouse Genome G4122A A_52_P432396 BB440691.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P432396 AK081765.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P432396 AK050824.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P432396 AK080302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P481140 BC048731.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P481140 AK081765.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P192569 AK081131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333020 AK154145.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P333020 AK041511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P419632 BC085144.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P419632 AK142089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P419632 AK154145.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P419632 AK169954.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P419632 AK051146.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P419632 BC085144.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P419632 AK142089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P419632 AK154145.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P419632 AK169954.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P419632 AK051146.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146758 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109907 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05921025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06439656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06361762 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506117 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05921017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06280186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06179847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506166 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05921024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506113 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06040809 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06530086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05921014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05921030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06622999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05921021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05921020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06623003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06331975 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06032790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06479126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06394589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06530092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06179846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06215103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06341677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25506156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25505944 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10557701 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557700 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557658 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557657 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557702 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557661 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557656 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557662 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557659 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557678 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568259 - -
          Affymetrix Mouse Gene 1.0 ST Array 10557660 - -
          Affymetrix HT MG-430 PM Array Plate 1457718_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1441653_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1437909_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1439978_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568258 - -
          Affymetrix Mouse Gene 1.1 ST Array 10557655 - -
          Mouse Genome 430 2.0 Array 1441653_PM_at - -
          Mouse Genome 430 2.0 Array 1439978_at - -
          Mouse Genome 430 2.0 Array 1439978_PM_at - -
          Mouse Genome 430 2.0 Array 1457718_PM_at - -
          Mouse Genome 430 2.0 Array 1441653_at - -
          Mouse Genome 430 2.0 Array 1437909_PM_at - -
          Mouse Genome 430 2.0 Array 1437909_at - -
          Mouse Genome 430 2.0 Array 1457718_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100043597_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100043597_PM_at - -
          Mouse Expression Set 430 1437909_PM_at - -
          Mouse Expression Set 430 1437909_at - -
          Mouse Expression Set 430 1441653_PM_at - -
          Mouse Expression Set 430 1457718_PM_at - -
          Mouse Expression Set 430 1441653_at - -
          Mouse Expression Set 430 1457718_at - -
          Mouse Expression Set 430 1439978_at - -
          Mouse Expression Set 430 1439978_PM_at - -
          Murine Genome U74v2 Set 99825_PM_at - -
          Murine Genome U74v2 Set 97163_g_at - -
          Murine Genome U74v2 Set 97163_PM_g_at - -
          Murine Genome U74v2 Set 99825_at - -
          Murine Genome U74v2 Set 135158_at - -
          Murine Genome U74v2 Set 134342_r_at - -
          Murine Genome U74v2 Set 135158_PM_at - -
          Murine Genome U74v2 Set 130336_PM_at - -
          Murine Genome U74v2 Set 130336_at - -
          Murine Genome U74v2 Set 134342_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1437909_at - -
          Mouse Genome 430A 2.0 Array 1437909_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1437909_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1437909_at - -
          Affymetrix HT Mouse Genome 430B Array 1457718_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441653_at - -
          Affymetrix HT Mouse Genome 430B Array 1457718_at - -
          Affymetrix HT Mouse Genome 430B Array 1441653_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439978_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1439978_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 885416 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline