Search
Search
| Gene Symbol: Tsr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164578.1 | NP_001158050.1 | 2 - 3 | 204 | 81 | |
| NM_175146.4 | NP_780355.2 | 2 - 3 | 204 | 81 | ||
| GenBank mRNA | AK044504.1 | - | 2 - 3 | 180 | 81 | |
| BC025008.1 | - | 2 - 3 | 167 | 81 | ||
| BC049668.1 | - | 2 - 3 | 186 | 6535058 | 81 | |
| CF732210.1 | - | 2 - 3 | 168 | 81 | ||
| CJ072126.1 | - | 2 - 3 | 206 | 81 | ||
Set Membership: |
|