| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P111714 |
BC005522.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P111714 |
BC024680.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P111714 |
BC048955.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P111714 |
AK047081.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_66_P111714 |
AB052760.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
4762 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
750900 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
690480 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
616291 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
404581 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
689852 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
902856 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
305818 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
773649 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
711406 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
204181 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
878123 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
518183 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
973580 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
54544 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
219206 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
510840 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
611036 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
926840 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
279627 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
626355 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
82396 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
522872 |
AK050798.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
522872 |
J03074.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
522872 |
NM_010465.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
522872 |
X16510.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
522872 |
X16511.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
521211 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
302233 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
692263 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
969458 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
748721 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
67213 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
306956 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
691141 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
666836 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
506236 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
256498 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
777340 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
468500 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
827667 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
227524 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
733082 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
750443 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
832623 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
535998 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
377374 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
417207 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
9104 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
845094 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
754006 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
227641 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
107989 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
171633 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
130272 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
10612 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
132538 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
126124 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
10388 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
157921 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
93838 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P130917 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P114799 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P111714 |
BC005522.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P111714 |
BC024680.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P111714 |
BC048955.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P111714 |
AK047081.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_66_P111714 |
AB052760.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07141948 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P130917 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07142179 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07142137 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07141991 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P114799 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P75755 |
AK047081.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P319425 |
BC005522.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P319425 |
BC024680.1 |
mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P07142137 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P114799 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P130917 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327723 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327691 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327351 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327411 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08958456 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327342 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327346 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327396 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327719 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08838460 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327412 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08480072 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327367 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327405 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327359 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327687 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327347 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327308 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08562650 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327301 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327326 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08640175 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08541947 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327681 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08925786 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327292 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327432 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327340 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327339 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327311 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327439 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327431 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327706 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327708 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327318 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327710 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327330 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08723983 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327307 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327707 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08229500 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08305501 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327685 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327419 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327438 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08519454 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327382 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08884350 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327446 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327671 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327690 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327366 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327281 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327345 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08541936 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327674 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327360 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327325 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327314 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327430 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327275 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327717 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327416 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327320 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327436 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327394 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08623269 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327398 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327357 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327424 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327678 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08723982 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327730 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327329 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08958450 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327709 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327727 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327331 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08640179 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08825797 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327305 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08489870 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327434 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327689 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08480070 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327442 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327423 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327362 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327354 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327288 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08376160 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327356 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327386 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327344 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08541945 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327276 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08562636 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327715 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327413 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327444 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327421 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327722 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08359013 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327279 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327695 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327392 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327677 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327390 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327290 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08376171 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327696 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327365 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327693 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327420 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327336 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327387 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327447 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327285 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327384 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327295 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327294 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327698 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327332 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327729 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327297 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327700 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08423568 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327363 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327403 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327672 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08480071 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327349 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327414 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08758237 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P27327425 |
- |
- |
| Agilent Six_ChIP_02_of_03_v2 |
A_68_P08305502 |
- |
- |