| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1900003678.mm.1 |
NM_028785.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003678.mm.1 |
NM_028785.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003678.mm.1 |
TC1900000399.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003676.mm.1 |
NM_028785.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003676.mm.1 |
NM_028785.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003676.mm.1 |
TC1900000399.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003677.mm.1 |
NM_028785.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003677.mm.1 |
NM_028785.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003677.mm.1 |
TC1900000399.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003675.mm.1 |
NM_028785.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003675.mm.1 |
NM_028785.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003675.mm.1 |
TC1900000399.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003673.mm.1 |
NM_028785.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003673.mm.1 |
NM_028785.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003673.mm.1 |
TC1900000399.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900003674.mm.1 |
NM_028785.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003674.mm.1 |
NM_028785.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900003674.mm.1 |
TC1900000399.mm.1 |
MTA TC |