| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10375888 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10375892 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10375887 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10375891 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10375889 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10375890 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1439782_PM_s_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1449656_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1419893_PM_at |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1439781_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10375886 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10385709 |
- |
- |
| Mouse Genome 430 2.0 Array |
1439781_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1419893_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1449656_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1419893_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1449656_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1439781_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1439782_PM_s_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1439782_s_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
97775_at |
- |
- |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array |
97775_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1100004479.mm.1 |
NR_027958.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004479.mm.1 |
NR_027958.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004479.mm.1 |
TC1100000574.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100004480.mm.1 |
NR_027958.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004480.mm.1 |
NR_027958.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100004480.mm.1 |
TC1100000574.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1439782_s_at |
- |
- |
| Mouse Expression Set 430 |
1439781_PM_at |
- |
- |
| Mouse Expression Set 430 |
1439781_at |
- |
- |
| Mouse Expression Set 430 |
1439782_PM_s_at |
- |
- |
| Murine Genome U74v2 Set |
104411_PM_at |
- |
- |
| Murine Genome U74v2 Set |
104411_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1419893_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1449656_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1419893_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1449656_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1439782_PM_s_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1439781_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1439781_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1439782_s_at |
- |
- |