Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00614265_m1
          See other NLRP4F GE Assays ›
          Gene Symbol:
          Nlrp4f
          Gene Name:
          NLR family, pyrin domain containing 4F
          Gene Aliases:
          C330026N02Rik, C86045, Nalp-kappa, Nalp4f
          Chromosome Location:
          Chr.13: 65177111 - 65205716 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00614265_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nlrp4f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175290.4 NP_780499.3 7 - 8 2635 61
          GenBank mRNA AK049352.1 - 5 - 6 1038 61
          BC098479.1 - 5 - 6 1057 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002211.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nlrp4f

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          biological_process
          nucleotide binding
          molecular_function
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102444 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102445 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102443 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102440 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102446 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102452 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102453 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102448 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102449 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102441 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102451 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102442 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102450 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102447 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955871 AY596201.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955871 BC098479.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955871 CD541102.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955871 NM_175290.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955871 AK049352.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428096 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 631917 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112958 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569436 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909930 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 7304 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79382 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130620 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163569 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168776 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955871 AY596201.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955871 BC098479.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955871 CD541102.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955871 NM_175290.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955871 AK049352.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29102314 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163569 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130620 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168776 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07902715 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02710767 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07902686 - -
          Whole Mouse Genome G4122A A_51_P154637 AY596201.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154637 BC098479.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154637 CD541102.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154637 NM_175290.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P154637 AK049352.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114314 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111586 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130620 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168776 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163569 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11008272 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10972908 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10546833 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11075117 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102446 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102445 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102444 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102449 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102448 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102442 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102450 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102441 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102452 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102451 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102443 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102440 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102447 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29102453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29102299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102282 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11097765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10668791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10668790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102309 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102276 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102334 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102319 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29102303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10615199 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10410190 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410189 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410198 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410197 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410202 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410201 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410193 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410196 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410188 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410200 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410192 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410191 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410195 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410203 - -
          Affymetrix HT MG-430 PM Array Plate 1436794_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10410187 - -
          Mouse Genome 430 2.0 Array 1436794_at - -
          Mouse Genome 430 2.0 Array 1436794_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 97895_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 97895_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300014314.mm.1 NM_175290.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014314.mm.1 NM_175290.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014314.mm.1 TC1300002211.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014312.mm.1 NM_175290.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014312.mm.1 NM_175290.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014312.mm.1 TC1300002211.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014315.mm.1 NM_175290.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014315.mm.1 NM_175290.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014315.mm.1 TC1300002211.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014311.mm.1 NM_175290.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014311.mm.1 NM_175290.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014311.mm.1 TC1300002211.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014313.mm.1 NM_175290.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014313.mm.1 NM_175290.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014313.mm.1 TC1300002211.mm.1 MTA TC
          Mouse Expression Set 430 1436794_PM_at - -
          Mouse Expression Set 430 1437299_PM_at - -
          Mouse Expression Set 430 1437299_at - -
          Mouse Expression Set 430 1436794_at - -
          Murine Genome U74v2 Set 137631_PM_f_at - -
          Murine Genome U74v2 Set 137631_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1436794_at - -
          Affymetrix HT Mouse Genome 430B Array 1436794_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 743248 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3610601 - -
          Illumina mouse-6 v1.0 expression beadchip 3610601 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003610601 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2601376 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2601376 - -
          mouse-6 v1.1 (Illumina) scl43813.10.1_83-S - -
          Illumina Sentrix mouse-6.v1 bead array 3610601 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43813.10.1_83-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3610601 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2601376 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2601376 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline