Search
Search

Zfp513| Gene Symbol: Zfp513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177901.1 | NP_001171372.1 | 1 - 2 | 462 | 79 | |
| NM_175311.4 | NP_780520.3 | 2 - 3 | 475 | 79 | ||
| GenBank mRNA | AK052459.1 | - | 1 - 2 | 307 | 79 | |
| AK154195.1 | - | 1 - 2 | 463 | 79 | ||
| BC058976.1 | - | 2 - 3 | 405 | 5685033 | 79 | |
Set Membership: |
|
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23577043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23577059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23577058 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23577042 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23577056 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077535 | BC058976.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077535 | NM_175311.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077535 | AK052459.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077535 | AK154195.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2077535 | NM_001177901.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 308027 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 360840 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822524 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P171378 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077535 | BC058976.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077535 | NM_175311.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077535 | AK052459.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077535 | AK154195.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2077535 | NM_001177901.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577058 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577037 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577040 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577041 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23577043 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P171378 | - | - |
| Whole Mouse Genome G4122A | A_51_P511529 | BC058976.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511529 | NM_175311.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511529 | AK052459.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511529 | AK154195.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P511529 | AK177145.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112454 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P171378 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04450817 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04450828 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04450775 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04450796 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04450829 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23577059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23577042 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23577056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23577043 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23577058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577040 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577015 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577013 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577005 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577036 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577041 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576969 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577012 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577043 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576999 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23576992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23577039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577057 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577030 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577033 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577049 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577047 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577038 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04450796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23577041 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10529138 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529137 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529134 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529139 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529141 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529135 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10529136 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437483_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10529133 | - | - |
| Mouse Genome 430 2.0 Array | 1437483_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437483_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101023_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101023_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500019907.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019907.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019907.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019907.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019907.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019911.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019911.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019911.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019911.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019911.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019915.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019915.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019915.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019915.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019915.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019922.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019922.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019922.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019922.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019922.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019930.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019930.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019930.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019909.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019909.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019909.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019909.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019909.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019919.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019919.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019919.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019919.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019919.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019926.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019926.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019926.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019926.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019926.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019913.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019913.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019913.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019913.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019913.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019921.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019921.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019921.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019921.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019921.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019906.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019906.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019906.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019906.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019906.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019914.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019914.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019914.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019914.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019914.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019917.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019917.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019917.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019917.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019917.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019923.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019923.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019923.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019923.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019923.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019927.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019927.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019927.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019924.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019924.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019924.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019908.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019908.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019908.mm.1 | NM_175311.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019908.mm.1 | NM_175311.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019908.mm.1 | TC0500002177.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500019925.mm.1 | NM_001177901.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019925.mm.1 | NM_001177901.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500019925.mm.1 | TC0500002177.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437483_at | - | - |
| Mouse Expression Set 430 | 1437483_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437483_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437483_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 446655 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4590594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 380673 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4590594 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004590594 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000380673 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2461853 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1232999 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1232999 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2461853 | - | - |
| mouse-6 v1.1 (Illumina) | scl0101023.1_246-S | - | - |
| mouse-6 v1.1 (Illumina) | scl26737.1.11_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4590594 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 380673 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0101023.1_246-S | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl26737.1.11_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4590594 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380673 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2461853 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2936604 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1257753 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2936604 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1257753 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2461853 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0101023.1_246-S | - | - |