Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00614486_m1
          See other PLEKHG4 GE Assays ›
          Gene Symbol:
          Plekhg4
          Gene Name:
          pleckstrin homology domain containing, family G (with RhoGef domain) member 4
          Gene Aliases:
          4931414L13Rik, AI448583, mFLJ00068
          Chromosome Location:
          Chr.8: 105369441 - 105382862 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00614486_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Plekhg4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081333.1 NP_001074802.1 13 - 14 2419 101
          GenBank mRNA AK029859.1 - 11 - 12 3091 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001217.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503883 BC024792.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503883 AK131125.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503883 NM_001081333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503883 AK029859.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503883 BC024792.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503883 AK131125.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503883 NM_001081333.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503883 AK029859.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 838528 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378508 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 553331 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393947 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163525 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503883 BC024792.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503883 AK131125.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503883 NM_001081333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503883 AK029859.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503883 BC024792.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503883 AK131125.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503883 NM_001081333.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503883 AK029859.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163525 - -
          Whole Mouse Genome G4122A A_51_P503883 BC024792.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P503883 AK131125.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P503883 NM_001081333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P503883 AK029859.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P503883 BC024792.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P503883 AK131125.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P503883 NM_001081333.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P503883 AK029859.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133430 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133434 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26133416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133460 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06822240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06920438 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06822275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133462 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06920447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133423 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06920478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07196359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06920444 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133435 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133430 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06727757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133385 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06822277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06720121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133390 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26133375 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574757 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574762 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574746 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574752 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574748 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574760 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574758 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574749 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574751 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574754 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574763 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574764 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574745 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574761 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574759 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574747 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574744 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574755 - -
          Affymetrix HT MG-430 PM Array Plate 1457145_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10574743 - -
          Mouse Genome 430 2.0 Array 1457145_PM_at - -
          Mouse Genome 430 2.0 Array 1457145_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102075_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 102075_at - -
          Affymetrix Mouse Clariom S Array PSR0800009629.mm.1 NM_001081333.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009629.mm.1 NM_001081333.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009629.mm.1 TC0800001217.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009628.mm.1 NM_001081333.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009628.mm.1 NM_001081333.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009628.mm.1 TC0800001217.mm.1 MTA TC
          Mouse Expression Set 430 1457145_at - -
          Mouse Expression Set 430 1457145_PM_at - -
          Murine Genome U74v2 Set 134679_at - -
          Murine Genome U74v2 Set 134679_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1457145_at - -
          Affymetrix HT Mouse Genome 430B Array 1457145_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 382035 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2726005 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2726005 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline