Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00614902_g1
          See other 4933402P03RIK GE Assays ›
          Gene Symbol:
          4933402P03Rik
          Gene Name:
          RIKEN cDNA 4933402P03 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 69816560 - 69818487 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00614902_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933402P03Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175368.1 NP_780577.1 1 - 2 181 117
          GenBank mRNA AK077090.1 - 1 - 2 209 117
          BC137961.1 - 1 - 2 170 8860980 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003067.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 4933402P03Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P593853 AK077090.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P593853 BC137961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P593853 NM_175368.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P593853 AK077090.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P593853 BC137961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P593853 NM_175368.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351538 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P104281 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P593853 AK077090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P593853 BC137961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P593853 NM_175368.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P593853 AK077090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P593853 BC137961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P593853 NM_175368.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27897283 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P104281 - -
          Whole Mouse Genome G4122A A_52_P593853 AK077090.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593853 BC137961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593853 NM_175368.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593853 AK077090.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593853 BC137961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P593853 NM_175368.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138060 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P104281 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08995410 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08995395 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08995406 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08995405 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08995394 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08995403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27897274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09439922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08995383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09439917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09045882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08995405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08995392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08995389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27897286 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10387654 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387653 - -
          Affymetrix Mouse Gene 1.0 ST Array 10387652 - -
          Affymetrix Mouse Gene 1.1 ST Array 10387651 - -
          Affymetrix Mouse Clariom S Array PSR1100028136.mm.1 NM_175368.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028136.mm.1 NM_175368.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028136.mm.1 TC1100003067.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028137.mm.1 NM_175368.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028137.mm.1 NM_175368.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028137.mm.1 TC1100003067.mm.1 MTA TC
          Murine Genome U74v2 Set 167664_i_at - -
          Murine Genome U74v2 Set 167664_PM_i_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 710850 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1190091 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190091 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254829 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254829 - -
          mouse-6 v1.1 (Illumina) scl40014.3.1_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190091 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40014.3.1_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190091 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1254829 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254829 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline