Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00615035_s1
          See other COA5 GE Assays ›
          Gene Symbol:
          Coa5
          Gene Name:
          cytochrome C oxidase assembly factor 5
          Gene Aliases:
          1700001A24Rik, 6330578E17Rik, Pet191
          Chromosome Location:
          Chr.1: 37417085 - 37430103 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00615035_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Coa5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198006.4 NP_932123.3 3 3547 89
          GenBank mRNA AK075747.1 - 1 1652 89
          AK166651.1 - 3 3476 89
          AK169639.1 - 3 3534 89
          BC069969.1 - 1 2102 89
          BC096450.1 - 1 2163 89
          BQ176168.1 - 1 89

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Coa5

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153982 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153981 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153983 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962035 BC069969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962035 BC096450.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962035 AK169639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962035 AK166651.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962035 AK075747.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437938 AK169639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437938 BC002137.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437938 AK166651.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437938 AK050432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437938 NM_198006.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437938 AK169639.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437938 BC002137.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437938 AK166651.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437938 AK050432.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P437938 NM_198006.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 396853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 786595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 705026 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 224701 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 129561 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944066 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 30499 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92355 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167636 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124123 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962035 BC069969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962035 BC096450.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962035 AK169639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962035 AK166651.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962035 AK075747.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437938 AK169639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437938 BC002137.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437938 AK166651.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437938 AK050432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437938 NM_198006.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437938 AK169639.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437938 BC002137.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437938 AK166651.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437938 AK050432.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P437938 NM_198006.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20153968 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124123 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167636 - -
          Whole Mouse Genome G4122A A_52_P670154 BC069969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P670154 BC096450.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P670154 AK169639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P670154 AK166651.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P670154 AK075747.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437938 AK169639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437938 BC002137.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437938 AK166651.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437938 AK050432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437938 NM_198006.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437938 AK169639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437938 BC002137.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437938 AK166651.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437938 AK050432.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P437938 NM_198006.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P330577 AK040907.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167636 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124123 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00204907 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01145217 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00611619 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01145218 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00611621 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00850416 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00204901 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00850412 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00943784 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00204894 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00650968 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153981 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153983 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20153982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20153946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01145217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00816933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20153965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20153970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20153977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20153967 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00611619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20153974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20153975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01145218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20153972 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10353997 - -
          Affymetrix Mouse Gene 1.0 ST Array 10354002 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353995 - -
          Affymetrix Mouse Gene 1.0 ST Array 10354001 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353994 - -
          Affymetrix Mouse Gene 1.0 ST Array 10354000 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353999 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353996 - -
          Affymetrix Mouse Gene 1.0 ST Array 10353998 - -
          Affymetrix HT MG-430 PM Array Plate 1426356_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1426355_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10353993 - -
          Mouse Genome 430 2.0 Array 1426356_PM_at - -
          Mouse Genome 430 2.0 Array 1426355_a_at - -
          Mouse Genome 430 2.0 Array 1426355_PM_a_at - -
          Mouse Genome 430 2.0 Array 1426356_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76178_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76178_at - -
          Affymetrix Mouse Clariom S Array PSR0100017688.mm.1 NM_198006.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017688.mm.1 NM_198006.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017688.mm.1 TC0100002203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017694.mm.1 NM_198006.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017694.mm.1 NM_198006.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017694.mm.1 TC0100002203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017690.mm.1 NM_198006.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017690.mm.1 NM_198006.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017690.mm.1 TC0100002203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017689.mm.1 NM_198006.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017689.mm.1 NM_198006.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017689.mm.1 TC0100002203.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100017692.mm.1 NM_198006.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017692.mm.1 NM_198006.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100017692.mm.1 TC0100002203.mm.1 MTA TC
          Mouse Expression Set 430 1426356_at - -
          Mouse Expression Set 430 1426356_PM_at - -
          Mouse Expression Set 430 1426355_a_at - -
          Mouse Expression Set 430 1426355_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1426356_at - -
          Mouse Genome 430A 2.0 Array 1426355_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1426355_a_at - -
          Mouse Genome 430A 2.0 Array 1426356_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426355_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1426356_at - -
          Affymetrix HT Mouse Genome 430A Array 1426356_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426355_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 308985 - -
          ABI Mouse Genome Survey Microarray 777663 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7100133 - -
          Illumina mouse-6 v1.0 expression beadchip 7100133 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2650106 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1248618 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2650106 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1248618 - -
          mouse-6 v1.1 (Illumina) scl076178.1_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 7100133 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl076178.1_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100133 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1248618 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1248618 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0