Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00615062_s1
          See other RBM15B GE Assays ›
          Gene Symbol:
          Rbm15b
          Gene Name:
          RNA binding motif protein 15B
          Gene Aliases:
          1810017N16Rik
          Chromosome Location:
          Chr.9: 106883985 - 106887000 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00615062_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbm15b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175402.4 NP_780611.3 1 1708 66
          GenBank mRNA AK075777.1 - 1 651 66
          BC052180.1 - 2 890 6332977 66
          BC078452.1 - 2 1531 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbm15b

          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of alternative mRNA splicing, via spliceosome
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          mRNA processing
          RNA splicing
          nucleotide binding
          nucleic acid binding
          RNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035584 AK137366.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035584 AK162618.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503192 BC052180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503192 BC057609.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503192 BC078452.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503192 AK075777.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503192 NM_175402.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503192 BC052180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503192 BC057609.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503192 BC078452.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503192 AK075777.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P503192 NM_175402.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 157295 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126238 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503192 BC052180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503192 BC057609.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503192 BC078452.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503192 AK075777.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503192 NM_175402.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503192 BC052180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503192 BC057609.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503192 BC078452.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503192 AK075777.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P503192 NM_175402.4 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02023633 - -
          Whole Mouse Genome G4122A A_51_P503194 AK075777.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10596574 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596571 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596573 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596572 - -
          Affymetrix HT MG-430 PM Array Plate 1454871_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1437778_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10596570 - -
          Mouse Genome 430 2.0 Array 1437778_at - -
          Mouse Genome 430 2.0 Array 1454871_PM_at - -
          Mouse Genome 430 2.0 Array 1454871_at - -
          Mouse Genome 430 2.0 Array 1437778_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900025141.mm.1 NM_175402.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025141.mm.1 NM_175402.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025141.mm.1 TC0900003021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025139.mm.1 NM_175402.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025139.mm.1 NM_175402.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025139.mm.1 TC0900003021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025142.mm.1 TC0900003021.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025140.mm.1 NM_175402.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025140.mm.1 NM_175402.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025140.mm.1 TC0900003021.mm.1 MTA TC
          Mouse Expression Set 430 1454798_at - -
          Mouse Expression Set 430 1454798_PM_at - -
          Mouse Expression Set 430 1454871_at - -
          Mouse Expression Set 430 1437778_at - -
          Mouse Expression Set 430 1437778_PM_at - -
          Mouse Expression Set 430 1454871_PM_at - -
          Mouse Genome 430A 2.0 Array 1454798_at - -
          Mouse Genome 430A 2.0 Array 1454798_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454871_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1437778_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454871_at - -
          Affymetrix HT Mouse Genome 430B Array 1437778_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 645493 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 460139 - -
          Illumina mouse-6 v1.0 expression beadchip 460139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000460139 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2743526 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2743526 - -
          mouse-6 v1.1 (Illumina) scl35383.2.97_43-S - -
          Illumina Sentrix mouse-6.v1 bead array 460139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35383.2.97_43-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460139 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2743524 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl35383.2.97_43-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline