Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00615375_m1
          See other ABRA GE Assays ›
          Gene Symbol:
          Abra
          Gene Name:
          actin-binding Rho activating protein
          Gene Aliases:
          C130068O12Rik, STARS
          Chromosome Location:
          Chr.15: 41865293 - 41869720 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00615375_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Abra
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175456.4 NP_780665.1 1 - 2 707 105
          GenBank mRNA AK045186.1 - 536 105
          AK081696.1 - 1 - 2 708 105
          AK138943.1 - 406 105
          BC119198.1 - 1 - 2 8733947 105
          BC120842.1 - 1 - 2 40129765 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500001446.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Abra

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transport
          protein transport
          positive regulation of Rho protein signal transduction
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of sequence-specific DNA binding transcription factor activity
          transcription coactivator activity
          actin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132070 AK138943.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132070 NM_175456.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132070 AK045186.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P132070 AK081696.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 745575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246709 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 665750 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71606 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P145451 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P132070 AK138943.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P132070 NM_175456.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P132070 AK045186.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P132070 AK081696.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167932 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30167920 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P145451 - -
          Whole Mouse Genome G4122A A_52_P348648 AK138943.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P348648 NM_175456.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P348648 AK045186.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P348648 AK081696.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132070 AK138943.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132070 NM_175456.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132070 AK045186.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132070 AK081696.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P145451 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11901148 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11901132 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11901151 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11901137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30167897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167879 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167970 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167889 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167963 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11901135 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11842332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167903 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12193575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167960 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167923 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11901152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167961 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12101753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11840278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167871 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30167946 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10428372 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428373 - -
          Affymetrix Mouse Gene 1.0 ST Array 10428371 - -
          Affymetrix HT MG-430 PM Array Plate 1458455_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10428370 - -
          Mouse Genome 430 2.0 Array 1458455_PM_at - -
          Mouse Genome 430 2.0 Array 1458455_at - -
          Affymetrix Mouse Clariom S Array PSR1500011083.mm.1 NM_175456.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500011083.mm.1 NM_175456.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500011083.mm.1 TC1500001446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500011084.mm.1 NM_175456.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500011084.mm.1 NM_175456.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500011084.mm.1 TC1500001446.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500011085.mm.1 NM_175456.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500011085.mm.1 NM_175456.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500011085.mm.1 TC1500001446.mm.1 MTA TC
          Mouse Expression Set 430 1458455_PM_at - -
          Mouse Expression Set 430 1458455_at - -
          Affymetrix HT Mouse Genome 430B Array 1458455_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458455_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 352567 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 100060279 - -
          Illumina mouse-6 v1.0 expression beadchip 730348 - -
          Illumina mouse-6 v1.0 expression beadchip 100060279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000730348 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2732401 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2732401 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2439909 - -
          mouse-6 v1.1 (Illumina) scl00223513.2_240-S - -
          mouse-6 v1.1 (Illumina) scl47257.2_377-S - -
          Illumina Sentrix mouse-6.v1 bead array 100060279 - -
          Illumina Sentrix mouse-6.v1 bead array 730348 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00223513.2_240-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 730348 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 100060279 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2732401 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2732401 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline