Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00615378_m1
          See other MROH1 GE Assays ›
          Gene Symbol:
          Mroh1
          Gene Name:
          maestro heat-like repeat family member 1
          Gene Aliases:
          D330001F17Rik, Heatr7a
          Chromosome Location:
          Chr.15: 76380467 - 76453039 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00615378_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mroh1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001162489.1 NP_001155961.1 3 - 4 321 74
          NM_175457.4 NP_780666.3 4 - 5 385 74
          GenBank mRNA AK036810.1 - 4 - 5 386 74
          AK038863.1 - 4 - 5 366 74
          AK052153.1 - 3 - 4 336 74
          AK090131.1 - 4 - 5 377 74
          AK147808.1 - 4 - 5 376 74
          AK147897.1 - 4 - 5 373 74
          AK170702.1 - 4 - 5 379 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000591.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mroh1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087885 AK170702.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087885 AK183494.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087885 BC039227.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087885 AK090131.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2087885 AK122562.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2210011 AK036810.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263966 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 417925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 746766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 231095 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118568 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 489310 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90373 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 168942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 850657 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 456842 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 941670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 850992 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 939712 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 789696 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 164319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907960 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 106167 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773523 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229513 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382844 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 933291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696846 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 138633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 822293 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107087 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448284 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 226288 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 863599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 438074 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808323 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 378334 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35351 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 763825 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905740 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18353 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380354 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 209738 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 78443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 351980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111271 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 23313 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 36781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 110341 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 64922 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 153861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78763 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153291 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107996 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087885 AK170702.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087885 AK183494.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087885 BC039227.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087885 AK090131.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2087885 AK122562.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2210011 AK036810.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03073930 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08422302 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08422189 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03073941 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107996 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08422130 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153291 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03073893 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03073876 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03073809 - -
          Whole Mouse Genome G4122A A_52_P118516 AK170702.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P118516 BC039227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P118516 AK090131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P118516 AK122562.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P118516 AK147808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249385 AK170702.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249385 BC039227.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249385 AK090131.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249385 AK052153.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P249385 NM_175457.4 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153291 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08422302 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107996 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10424861 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424883 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424886 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424887 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424884 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424888 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424901 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424880 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424898 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424902 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424899 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424876 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424903 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424904 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424875 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424881 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424870 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424896 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424878 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424879 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424900 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424885 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424890 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424872 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424897 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424895 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424866 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424877 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424874 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424893 - -
          Affymetrix HT MG-430 PM Array Plate 1459218_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433808_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10424860 - -
          Affymetrix Mouse Gene 1.1 ST Array 10424894 - -
          Mouse Genome 430 2.0 Array 1433808_PM_at - -
          Mouse Genome 430 2.0 Array 1459218_PM_at - -
          Mouse Genome 430 2.0 Array 1459218_at - -
          Mouse Genome 430 2.0 Array 1433808_at - -
          Affymetrix Mouse Clariom S Array PSR1500004157.mm.1 NM_001162489.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004157.mm.1 NM_001162489.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004157.mm.1 NM_175457.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004157.mm.1 NM_175457.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004157.mm.1 TC1500000591.mm.1 MTA TC
          Mouse Expression Set 430 1433808_at - -
          Mouse Expression Set 430 1433808_PM_at - -
          Mouse Expression Set 430 1459218_at - -
          Mouse Expression Set 430 1459218_PM_at - -
          Murine Genome U74v2 Set 104122_PM_at - -
          Murine Genome U74v2 Set 104122_at - -
          Murine Genome U74v2 Set 162350_PM_at - -
          Murine Genome U74v2 Set 162350_at - -
          Mouse Genome 430A 2.0 Array 1433808_at - -
          Mouse Genome 430A 2.0 Array 1433808_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1433808_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1433808_at - -
          Affymetrix HT Mouse Genome 430B Array 1459218_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1459218_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 463889 - -
          ABI Mouse Genome Survey Microarray 509874 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107040402 - -
          Illumina mouse-6 v1.0 expression beadchip 107040402 - -
          Illumina mouse-6 v1.0 expression beadchip 104730632 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1230718 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2564561 - -
          Illumina Sentrix mouse-6.v1 bead array 107040402 - -
          Illumina Sentrix mouse-6.v1 bead array 104730632 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104730632 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 107040402 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2564564 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230718 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline