Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00615581_s1
          See other ACTBL2 GE Assays ›
          Gene Symbol:
          Actbl2
          Gene Name:
          actin, beta-like 2
          Gene Aliases:
          4732495G21Rik
          Chromosome Location:
          Chr.13: 111255013 - 111257749 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00615581_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Actbl2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175497.3 NP_780706.1 1 2144 75
          GenBank mRNA AK029110.1 - 1 2145 75
          BC111904.1 - 1 2110 40062272 75
          BC112429.1 - 1 2110 40062276 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Actbl2

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          nucleotide binding
          molecular_function
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162379 AK029110.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2162379 NM_175497.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 604094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547670 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 22716 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115230 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162379 AK029110.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2162379 NM_175497.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328012 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328004 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327989 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328026 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327992 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328001 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327980 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327995 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327984 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327991 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328007 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29327987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29328003 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115230 - -
          Whole Mouse Genome G4122A A_51_P395281 BC112429.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395281 BC111904.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395281 AK029110.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395281 AK029130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P395281 NM_175497.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P134550 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115230 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10818794 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10818779 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10818760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29328007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29327984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10506552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10818764 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10681976 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327952 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328027 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10534216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10815126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328102 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11046727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10938084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328024 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10681985 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11046735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327997 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327967 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328003 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328041 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327990 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10818753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327989 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10818795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328039 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10536508 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10797315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327982 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10681972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10938088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11004124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10667820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328051 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10815129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328005 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29327944 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29328050 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10407144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407143 - -
          Affymetrix Mouse Gene 1.1 ST Array 10407141 - -
          Affymetrix Mouse Clariom S Array PSR1300007884.mm.1 NM_175497.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007884.mm.1 NM_175497.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007884.mm.1 TC1300001248.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300007883.mm.1 NM_175497.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007883.mm.1 NM_175497.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007883.mm.1 TC1300001248.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300007882.mm.1 NM_175497.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007882.mm.1 NM_175497.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300007882.mm.1 TC1300001248.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 385907 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1660687 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660687 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2596812 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2596812 - -
          mouse-6 v1.1 (Illumina) scl44384.1.2_152-S - -
          Illumina Sentrix mouse-6.v1 bead array 1660687 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44384.1.2_152-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660687 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2596812 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2596812 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0