Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00615833_m1
          See other NLRP10 GE Assays ›
          Gene Symbol:
          Nlrp10
          Gene Name:
          NLR family, pyrin domain containing 10
          Gene Aliases:
          6430548I20Rik, Nalp10, Napl10, PAN5, Pynod
          Chromosome Location:
          Chr.7: 108921853 - 108930158 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00615833_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nlrp10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175532.3 NP_780741.1 2 - 3 331 73
          GenBank mRNA AK028575.1 - 2 - 3 354 73
          AK028802.1 - 2 - 3 333 73
          AK032446.1 - 2 - 3 327 73
          AY355344.1 - 1 - 2 300 73
          AY489193.1 - 1 - 2 284 73
          BC113194.1 - 2 - 3 327 40062457 73
          BC113801.1 - 2 - 3 327 40062458 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004065.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nlrp10

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          positive regulation of T-helper 1 type immune response
          defense response
          helper T cell enhancement of adaptive immune response
          dendritic cell migration
          innate immune response
          defense response to Gram-negative bacterium
          defense response to fungus
          positive regulation of T-helper 17 type immune response
          positive regulation of interleukin-8 secretion
          positive regulation of interleukin-6 secretion
          nucleotide binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P590625 AK028802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P590625 NM_175532.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P590625 AK028802.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P590625 NM_175532.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713974 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 27902 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 263227 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713372 AK133542.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713372 BC048372.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713372 NM_001033453.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713372 NM_001098230.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713372 NM_001098231.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33148 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142229 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P590625 AK028802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P590625 NM_175532.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P590625 AK028802.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P590625 NM_175532.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25405066 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06316622 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P142229 - -
          Whole Mouse Genome G4122A A_52_P590625 AK028802.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P590625 NM_175532.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P590625 AK028802.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P590625 NM_175532.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358825 AY355344.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P358825 AK032446.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109490 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P142229 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05925982 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05925983 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05918613 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06134262 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05918612 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06134263 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06134268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25405037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06302239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06568875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06302238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06568876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06061313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25405023 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566711 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566713 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566712 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566710 - -
          Affymetrix HT MG-430 PM Array Plate 1456211_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10566709 - -
          Mouse Genome 430 2.0 Array 1456211_PM_at - -
          Mouse Genome 430 2.0 Array 1456211_at - -
          Affymetrix Mouse Clariom S Array PSR0700033569.mm.1 NM_175532.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033569.mm.1 NM_175532.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033569.mm.1 TC0700004065.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033568.mm.1 NM_175532.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033568.mm.1 NM_175532.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033568.mm.1 TC0700004065.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033571.mm.1 NM_175532.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033571.mm.1 NM_175532.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033571.mm.1 TC0700004065.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700033570.mm.1 NM_175532.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033570.mm.1 NM_175532.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033570.mm.1 TC0700004065.mm.1 MTA TC
          Mouse Expression Set 430 1456211_PM_at - -
          Mouse Expression Set 430 1456211_at - -
          Affymetrix HT Mouse Genome 430B Array 1456211_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456211_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 692763 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4060519 - -
          Illumina mouse-6 v1.0 expression beadchip 104280131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004060519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2667249 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2453918 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2667249 - -
          mouse-6 v1.1 (Illumina) scl00244202.2_30-S - -
          mouse-6 v1.1 (Illumina) scl30839.3_120-S - -
          Illumina Sentrix mouse-6.v1 bead array 4060519 - -
          Illumina Sentrix mouse-6.v1 bead array 104280131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00244202.2_30-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4060519 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104280131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255593 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255593 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2453918 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00244202.2_30-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline