| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700017647.mm.1 |
TC0700002008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700017647.mm.1 |
NM_001128146.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017647.mm.1 |
NM_001128146.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017649.mm.1 |
TC0700002008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700017649.mm.1 |
NM_001128145.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017649.mm.1 |
NM_001128145.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017649.mm.1 |
NM_001128146.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017649.mm.1 |
NM_001128146.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017649.mm.1 |
NM_175533.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017649.mm.1 |
NM_175533.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017654.mm.1 |
TC0700002008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700017654.mm.1 |
NM_175533.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017654.mm.1 |
NM_175533.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017651.mm.1 |
TC0700002008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700017651.mm.1 |
NM_001128145.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017651.mm.1 |
NM_001128145.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017651.mm.1 |
NM_001128146.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017651.mm.1 |
NM_001128146.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017651.mm.1 |
NM_175533.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017651.mm.1 |
NM_175533.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017656.mm.1 |
TC0700002008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700017656.mm.1 |
NM_175533.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017656.mm.1 |
NM_175533.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017650.mm.1 |
TC0700002008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700017650.mm.1 |
NM_175533.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017650.mm.1 |
NM_175533.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017652.mm.1 |
TC0700002008.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700017652.mm.1 |
NM_175533.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700017652.mm.1 |
NM_175533.3 |
Refseq |