Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00615866_m1
          See other NIM1K GE Assays ›
          Gene Symbol:
          Nim1k
          Gene Name:
          NIM1 serine/threonine protein kinase
          Gene Aliases:
          E130304F04Rik, Nim1
          Chromosome Location:
          Chr.13: 119710094 - 119755882 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00615866_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nim1k
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175538.3 NP_780747.1 3 - 4 1404 119
          GenBank mRNA AK044462.1 - 2 - 3 1109 119
          AK047794.1 - 3 - 4 1394 119
          AK048694.1 - 3 - 4 1405 119
          AK053734.1 - 3 - 4 1411 119
          BC051189.1 - 2 - 3 1071 6491737 119
          BC106122.1 - 2 - 3 1098 4505882 119
          BC117955.1 - 3 - 4 1330 40098149 119
          BC117956.1 - 3 - 4 1328 40098152 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002747.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nim1k

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          phosphorylation
          intracellular signal transduction
          nucleotide binding
          magnesium ion binding
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057127 AK048694.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2057127 NM_175538.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057127 AK048694.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2057127 NM_175538.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334301 BC117956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334301 BC117955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334301 AK047794.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334301 AK048694.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334301 AK053734.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334542 BC117956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334542 BC117955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334542 AK048694.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P334542 NM_175538.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173459 BC117956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173459 BC106122.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173459 BC117955.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173459 AK047794.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P173459 AK048694.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137070 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137385 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106187 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10412436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10412442 - -
          Affymetrix HT MG-430 PM Array Plate 1434297_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10412435 - -
          Mouse Genome 430 2.0 Array 1434297_PM_at - -
          Mouse Genome 430 2.0 Array 1434297_at - -
          Affymetrix Mouse Clariom S Array PSR1300018576.mm.1 NM_175538.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018576.mm.1 NM_175538.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018576.mm.1 TC1300002747.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300018577.mm.1 NM_175538.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018577.mm.1 NM_175538.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018577.mm.1 TC1300002747.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300018579.mm.1 NM_175538.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018579.mm.1 NM_175538.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300018579.mm.1 TC1300002747.mm.1 MTA TC
          Mouse Expression Set 430 1434297_PM_at - -
          Mouse Expression Set 430 1434297_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 545241 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 110112 - -
          Illumina mouse-6 v1.0 expression beadchip 110112 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000110112 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2623315 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2623315 - -
          mouse-6 v1.1 (Illumina) scl00245269.1_243-S - -
          Illumina Sentrix mouse-6.v1 bead array 110112 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00245269.1_243-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 110112 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2623315 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2623315 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline