| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P517525 |
AK043681.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P517525 |
AK043681.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
691273 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
632859 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
580470 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
563713 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
896659 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
127636 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
290081 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
681419 |
AK137912.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
681419 |
AK157199.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
681419 |
NM_001033978.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
397398 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
761081 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
60974 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
645258 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
90233 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
88602 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P125553 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P123055 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P517525 |
AK043681.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P517525 |
AK043681.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335912 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335904 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335898 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335897 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335914 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335917 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335919 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335900 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335908 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335893 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335990 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335894 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335989 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335902 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335916 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335988 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335909 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335895 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335992 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335991 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335906 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P20335901 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00100290 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P123055 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P125553 |
- |
- |
| Whole Mouse Genome G4122A |
A_51_P517525 |
AK043681.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P517525 |
AK043681.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P134494 |
- |
- |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P104834 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P00100290 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P125553 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P123055 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00722354 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00722361 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P00722356 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336036 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335994 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335953 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335921 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336011 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335912 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335999 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335951 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335992 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335925 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335991 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336029 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336020 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335983 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335908 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335969 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335988 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335929 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335977 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335972 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335898 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335979 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335897 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336030 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335927 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335942 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335985 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335926 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336013 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335981 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335906 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335914 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335902 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336017 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335989 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336016 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336003 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336006 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335894 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336002 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335909 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335905 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335895 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335915 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335959 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335974 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335893 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335976 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336032 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335963 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335941 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336000 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335961 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335940 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335980 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336024 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336034 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336040 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335920 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20335962 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P20336023 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00722360 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335917 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00265811 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01023255 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P01023253 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00780889 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335897 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335927 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335925 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335902 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335978 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335924 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335903 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335976 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00170420 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335923 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335991 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335926 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00351859 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335983 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335992 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00265809 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335901 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00479726 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335904 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335988 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335898 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00479717 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335981 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00722357 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00351860 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335908 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335900 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00479715 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00805819 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335909 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335929 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335977 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20335920 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00862307 |
- |
- |