Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00616290_m1
          See other ESRP2 GE Assays ›
          Gene Symbol:
          Esrp2
          Gene Name:
          epithelial splicing regulatory protein 2
          Gene Aliases:
          9530027K23Rik, Rbm35b
          Chromosome Location:
          Chr.8: 106130403 - 106137528 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00616290_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Esrp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176838.2 NP_789808.1 2 - 3 538 68
          GenBank mRNA AK088755.1 - 2 - 3 407 68
          AK168488.1 - 2 - 3 386 68
          BC031444.1 - 2 - 3 538 4222237 68
          BC096581.1 - 2 - 3 378 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002852.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Esrp2

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          RNA splicing
          regulation of RNA splicing
          nucleotide binding
          nucleic acid binding
          RNA binding
          mRNA binding
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137937 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137939 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137938 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137942 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137944 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137948 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137946 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137940 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137936 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137945 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013143 BC031444.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013143 AK020578.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013143 AK088755.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013143 AK168488.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013143 NM_176838.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 669674 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 73086 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511213 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339921 AF210700.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339921 AK007321.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339921 AK029082.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339921 AK143807.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339921 BC010805.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339921 BC020159.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 339921 NM_026907.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 112165 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169608 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013143 BC031444.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013143 AK020578.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013143 AK088755.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013143 AK168488.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013143 NM_176838.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P296517 AK088755.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P296517 AK168488.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137936 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137925 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137928 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137930 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26137940 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169608 - -
          Whole Mouse Genome G4122A A_51_P296517 AK088755.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P296517 AK168488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P322301 BC031444.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P322301 AK020578.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P322301 AK088755.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P322301 AK168488.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P322301 NM_176838.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119853 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131212 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169608 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07134619 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07134630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07049600 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07134631 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07134633 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137937 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137939 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137940 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137946 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137944 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137945 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137938 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137936 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137942 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26137948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26137916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07049653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07134637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07049647 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07049640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137927 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07134633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07264263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07049635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137910 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137963 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07049632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137901 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06867329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07134631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137903 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26137929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06867327 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10581460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581465 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581471 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581462 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581458 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581457 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581466 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581468 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581463 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581467 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581469 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581464 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581470 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581472 - -
          Affymetrix Mouse Gene 1.0 ST Array 10581459 - -
          Affymetrix HT MG-430 PM Array Plate 1433683_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419977_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10581455 - -
          Mouse Genome 430 2.0 Array 1419977_PM_s_at - -
          Mouse Genome 430 2.0 Array 1433683_at - -
          Mouse Genome 430 2.0 Array 1419977_s_at - -
          Mouse Genome 430 2.0 Array 1433683_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800022018.mm.1 NM_176838.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022018.mm.1 NM_176838.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800022018.mm.1 TC0800002852.mm.1 MTA TC
          Mouse Expression Set 430 1433683_PM_at - -
          Mouse Expression Set 430 1433683_at - -
          Murine Genome U74v2 Set 161162_PM_at - -
          Murine Genome U74v2 Set 161162_at - -
          Mouse Genome 430A 2.0 Array 1433683_PM_at - -
          Mouse Genome 430A 2.0 Array 1433683_at - -
          Affymetrix HT Mouse Genome 430A Array 1419977_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1433683_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1433683_at - -
          Affymetrix HT Mouse Genome 430A Array 1419977_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 608983 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 106420072 - -
          Illumina mouse-6 v1.0 expression beadchip 520079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000520079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2603934 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1243939 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2603934 - -
          mouse-6 v1.1 (Illumina) scl34378.13_481-S - -
          mouse-6 v1.1 (Illumina) scl0077411.2_255-S - -
          Illumina Sentrix mouse-6.v1 bead array 106420072 - -
          Illumina Sentrix mouse-6.v1 bead array 520079 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0077411.2_255-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106420072 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 520079 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1224254 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224254 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1243939 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline