Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00616462_m1
          See other DNAH1 GE Assays ›
          Gene Symbol:
          Dnah1
          Gene Name:
          dynein, axonemal, heavy chain 1
          Gene Aliases:
          B230373P09Rik, Dnahc1, E030034C22Rik, MDHC7
          Chromosome Location:
          Chr.14: 31260375 - 31324063 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00616462_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dnah1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033668.1 NP_001028840.1 77 - 78 12698 60
          GenBank mRNA AK052631.1 - 19 - 20 3325 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001674.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          cytoskeletal protein microtubule or microtubule-binding cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial cilium movement
          sperm motility
          cilium movement involved in cell motility
          microtubule motor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2277970 AK163552.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2277970 AK052631.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2277970 NM_001033668.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2277970 AK046346.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108468 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164312 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2277970 AK163552.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2277970 AK052631.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2277970 NM_001033668.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2277970 AK046346.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P235174 Z83815.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164312 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08072513 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108468 - -
          Whole Mouse Genome G4122A A_51_P235174 Z83815.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P113861 Z83815.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P113861 NM_001033668.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128640 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112652 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108468 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511029 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511156 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11749683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11126009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11808881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11749665 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511148 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511149 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11535684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29510995 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11535681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29510996 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11535689 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11795733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11126013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11808873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11258538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11535695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511201 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11322493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511025 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11535672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11789472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511026 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11808872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511006 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29510992 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511137 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11576550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11126018 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511143 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511035 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511139 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11576545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29511189 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10418661 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418664 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418668 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418673 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418662 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418680 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418679 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418684 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418667 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418681 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418656 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418672 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418676 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418666 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418675 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418678 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418657 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418677 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418660 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418669 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418663 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418665 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418659 - -
          Affymetrix HT MG-430 PM Array Plate 1434594_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425307_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10418616 - -
          Mouse Genome 430 2.0 Array 1434594_at - -
          Mouse Genome 430 2.0 Array 1434594_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 110084_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 110084_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1400012880.mm.1 NM_001033668.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012880.mm.1 NM_001033668.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012880.mm.1 TC1400001674.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400012879.mm.1 NM_001033668.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012879.mm.1 NM_001033668.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012879.mm.1 TC1400001674.mm.1 MTA TC
          Mouse Expression Set 430 1434594_at - -
          Mouse Expression Set 430 1434594_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425307_at - -
          Affymetrix HT Mouse Genome 430A Array 1425307_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434594_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434594_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 450365 - -
          ABI Mouse Genome Survey Microarray 907648 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2715311 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1238791 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2715311 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2429227 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238791 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline