Search
Search

Trpm3| Gene Symbol: Trpm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035239.2 | NP_001030316.1 | 7 - 8 | 1154 | 71 | |
| NM_001035240.2 | NP_001030317.1 | 7 - 8 | 1154 | 71 | ||
| NM_001035241.2 | NP_001030318.1 | 7 - 8 | 1154 | 71 | ||
| NM_001035242.1 | NP_001030319.1 | 7 - 8 | 1154 | 71 | ||
| NM_001035243.2 | NP_001030320.1 | 7 - 8 | 1154 | 71 | ||
| NM_001035244.1 | NP_001030321.1 | 8 - 9 | 1277 | 71 | ||
| NM_177341.4 | NP_796315.2 | 7 - 8 | 1221 | 71 | ||
| GenBank mRNA | AJ544532.1 | - | 7 - 8 | 1154 | 71 | |
| AJ544533.1 | - | 7 - 8 | 1154 | 71 | ||
| AJ544534.1 | - | 7 - 8 | 1154 | 71 | ||
| AJ544535.1 | - | 7 - 8 | 1154 | 71 | ||
| AJ544536.1 | - | 7 - 8 | 1154 | 71 | ||
| AK044454.1 | - | 7 - 8 | 1223 | 71 | ||
| AK046908.1 | - | 8 - 9 | 1110 | 71 | ||
| AK051867.1 | - | 1 - 2 | 177 | 71 | ||
| AK053673.1 | - | 7 - 8 | 1039 | 71 | ||
| AK142504.1 | - | 8 - 9 | 1110 | 71 | ||
| AK173218.1 | - | 7 - 8 | 1225 | 71 | ||
| BC141400.1 | - | 7 - 8 | 1197 | 9056013 | 71 | |
| BC141412.1 | - | 7 - 8 | 1197 | 9056025 | 71 | |
| HM064407.1 | - | 7 - 8 | 1246 | 71 | ||
| HM064408.1 | - | 7 - 8 | 1246 | 71 | ||
| HM064409.1 | - | 7 - 8 | 1246 | 71 | ||
| HM064410.1 | - | 8 - 9 | 1321 | 71 | ||
| HM064411.1 | - | 7 - 8 | 1246 | 71 | ||
| JF706722.1 | - | 7 - 8 | 1217 | 71 | ||
| JX644970.1 | - | 7 - 8 | 1246 | 71 | ||
| JX644971.1 | - | 7 - 8 | 1246 | 71 | ||
| JX644972.1 | - | 7 - 8 | 1246 | 71 | ||
| JX644973.1 | - | 7 - 8 | 1246 | 71 | ||
| JX644974.1 | - | 7 - 8 | 1246 | 71 | ||
| JX644975.1 | - | 8 - 9 | 1321 | 71 | ||
| JX644976.1 | - | 7 - 8 | 1246 | 71 | ||
| JX644977.1 | - | 7 - 8 | 1246 | 71 | ||
| JX644978.1 | - | 7 - 8 | 1246 | 71 | ||
| JX644979.1 | - | 8 - 9 | 1321 | 71 | ||
| JX644980.1 | - | 8 - 9 | 1321 | 71 | ||
| JX644981.1 | - | 8 - 9 | 1321 | 71 | ||
| JX644982.1 | - | 8 - 9 | 1321 | 71 | ||
| JX644983.1 | - | 7 - 8 | 1246 | 71 | ||
| JX644984.1 | - | 7 - 8 | 1246 | 71 | ||
| JX644985.1 | - | 7 - 8 | 1246 | 71 | ||
| JX644986.1 | - | 7 - 8 | 1246 | 71 | ||
Set Membership: |
|