Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00616604_m1
          See other LAMP3 GE Assays ›
          Gene Symbol:
          Lamp3
          Gene Name:
          lysosomal-associated membrane protein 3
          Gene Aliases:
          1200002D17Rik, Cd208, DC-LAMP, DCLAMP, LAMP, TSC403
          Chromosome Location:
          Chr.16: 19653381 - 19706365 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00616604_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lamp3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177356.3 NP_796330.2 5 - 6 1204 76
          GenBank mRNA AJ510130.1 - 5 - 6 1123 76
          AK028039.1 - 5 - 6 1156 76
          AK033379.1 - 5 - 6 1157 76

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lamp3

          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091741 AK028039.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091741 AK033379.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091741 NM_177356.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091741 AJ510130.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 848549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 793319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970699 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335233 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 698075 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 786675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 766944 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 515265 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 398321 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 777314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311937 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81274 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 91633 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 782388 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131580 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117098 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13594 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153458 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152799 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091741 AK028039.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091741 AK033379.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091741 NM_177356.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091741 AJ510130.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P142807 AK028039.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P142807 AK033379.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P142807 NM_177356.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P142807 AJ510130.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P104593 AK028039.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P104593 AK033379.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P104593 NM_177356.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P104593 AJ510130.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239376 AK028039.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239376 AK033379.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239376 NM_177356.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P239376 AJ510130.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30584625 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08525608 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08525604 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08525584 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153458 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08525622 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152799 - -
          Whole Mouse Genome G4122A A_52_P104593 AK028039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P104593 AK033379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P104593 NM_177356.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P104593 AJ510130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P54647 AK164915.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P54647 AK086898.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P142807 AK028039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P142807 AK033379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P142807 NM_177356.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P142807 AJ510130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239376 AK028039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239376 AK033379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239376 NM_177356.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P239376 AJ510130.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115568 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100059 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152799 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153458 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08525608 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12657460 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12657463 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12657451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30584561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12657463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12959878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12578477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584584 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584619 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584591 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584627 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12649957 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584624 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12485020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584634 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12909585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30584607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12657457 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10438437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10438440 - -
          Affymetrix Mouse Gene 1.1 ST Array 10438435 - -
          Mouse Expression Set 430 1417057_PM_a_at - -
          Mouse Expression Set 430 1417057_a_at - -
          Murine Genome U74v2 Set 166940_PM_r_at - -
          Murine Genome U74v2 Set 166940_r_at - -
          Mouse Genome 430A 2.0 Array 1417057_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1417057_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 797969 - -
          ABI Mouse Genome Survey Microarray 901753 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 610059 - -
          MouseWG-6 v2 Expression BeadChip 1170711 - -
          Illumina mouse-6 v1.0 expression beadchip 2900154 - -
          Illumina mouse-6 v1.0 expression beadchip 610059 - -
          Illumina mouse-6 v1.0 expression beadchip 1170711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610059 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002900154 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001170711 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2745680 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2738854 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2745667 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2738854 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2745667 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2745680 - -
          mouse-6 v1.1 (Illumina) scl00239739.2_2-S - -
          mouse-6 v1.1 (Illumina) scl0239739.2_181-S - -
          mouse-6 v1.1 (Illumina) scl00239739.2_234-S - -
          Illumina Sentrix mouse-6.v1 bead array 1170711 - -
          Illumina Sentrix mouse-6.v1 bead array 2900154 - -
          Illumina Sentrix mouse-6.v1 bead array 610059 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0239739.2_181-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00239739.2_234-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00239739.2_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610059 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1170711 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2900154 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2745667 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2745680 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2738854 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2745667 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2745680 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2738854 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline