Search
Search

Sfmbt2| Gene Symbol: Sfmbt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198808.1 | NP_001185737.1 | 4 - 5 | 395 | 58 | |
| NM_001198809.1 | NP_001185738.1 | 4 - 5 | 474 | 58 | ||
| NM_177386.5 | NP_796360.2 | 5 - 6 | 518 | 58 | ||
| GenBank mRNA | AK040600.1 | - | 4 - 5 | 475 | 58 | |
| AK049505.1 | - | 4 - 5 | 337 | 58 | ||
| AK052339.1 | - | 4 - 5 | 461 | 58 | ||
| AK147557.1 | - | 4 - 5 | 400 | 58 | ||
| AK147651.1 | - | 5 - 6 | 373 | 58 | ||
| AK168444.1 | - | 6 - 7 | 423 | 58 | ||
| AK220408.1 | - | 2 - 3 | 260 | 58 | ||
| BB636293.1 | - | 4 - 5 | 475 | 58 | ||
| BC115510.1 | - | 4 - 5 | 329 | 58 | ||
| BC115511.1 | - | 4 - 5 | 329 | 58 | ||
| BC139354.1 | - | 4 - 5 | 306 | 8862376 | 58 | |
| BC145610.1 | - | 3 - 4 | 262 | 9054176 | 58 | |
Set Membership: |
|