Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617030_m1
          See other FBXW13 GE Assays ›
          Gene Symbol:
          Fbxw13
          Gene Name:
          F-box and WD-40 domain protein 13
          Gene Aliases:
          E330017E16
          Chromosome Location:
          Chr.9: 109179227 - 109195975 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617030_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxw13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177598.3 NP_808266.1 7 - 8 1024 116
          GenBank mRNA AK054339.1 - 7 - 8 1025 116
          BC119542.1 - 7 - 8 1011 40056515 116
          BC127237.1 - 6 - 7 847 40056507 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003078.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxw13

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P353541 BC127237.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P353541 BC119542.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P353541 NM_177598.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P353541 AK054339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P353541 BC127237.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P353541 BC119542.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P353541 NM_177598.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P353541 AK054339.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759299 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 122748 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 694208 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3591 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172603 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 172988 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20273 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169795 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144744 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P353541 BC127237.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P353541 BC119542.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P353541 NM_177598.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P353541 AK054339.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P353541 BC127237.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P353541 BC119542.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P353541 NM_177598.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P353541 AK054339.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26818807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26818797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26818799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26818785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26818811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26818812 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02027680 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P169795 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144744 - -
          Whole Mouse Genome G4122A A_52_P353541 BC127237.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353541 BC119542.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353541 NM_177598.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353541 AK054339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353541 BC127237.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353541 BC119542.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353541 NM_177598.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P353541 AK054339.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127002 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144744 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P169795 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08152248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07477399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08010971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08185643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07821012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08047790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07561425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08002148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07477401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818799 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10597007 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597005 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597009 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597003 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597001 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597008 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597010 - -
          Affymetrix Mouse Gene 1.0 ST Array 10597002 - -
          Affymetrix HT MG-430 PM Array Plate 1443298_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10597000 - -
          Mouse Genome 430 2.0 Array 1443298_at - -
          Mouse Genome 430 2.0 Array 1443298_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 211305_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 211305_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900025951.mm.1 NM_177598.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025951.mm.1 NM_177598.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025951.mm.1 TC0900003078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025954.mm.1 NM_177598.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025954.mm.1 NM_177598.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025954.mm.1 TC0900003078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025957.mm.1 NM_177598.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025957.mm.1 NM_177598.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025957.mm.1 TC0900003078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025955.mm.1 NM_177598.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025955.mm.1 NM_177598.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025955.mm.1 TC0900003078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025958.mm.1 NM_177598.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025958.mm.1 NM_177598.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025958.mm.1 TC0900003078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025952.mm.1 NM_177598.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025952.mm.1 NM_177598.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025952.mm.1 TC0900003078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025959.mm.1 NM_177598.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025959.mm.1 NM_177598.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025959.mm.1 TC0900003078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025961.mm.1 NM_177598.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025961.mm.1 NM_177598.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025961.mm.1 TC0900003078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025950.mm.1 NM_177598.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025950.mm.1 NM_177598.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025950.mm.1 TC0900003078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025953.mm.1 NM_177598.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025953.mm.1 NM_177598.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025953.mm.1 TC0900003078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025956.mm.1 NM_177598.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025956.mm.1 NM_177598.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025956.mm.1 TC0900003078.mm.1 MTA TC
          Mouse Expression Set 430 1443298_at - -
          Mouse Expression Set 430 1443298_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443298_at - -
          Affymetrix HT Mouse Genome 430B Array 1443298_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 503256 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3140338 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003140338 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2625152 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2625152 - -
          mouse-6 v1.1 (Illumina) scl35339.10.1_39-S - -
          Illumina Sentrix mouse-6.v1 bead array 3140338 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35339.10.1_39-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3140338 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2625152 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2625152 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline