Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617049_s1
          See other TMEM60 GE Assays ›
          Gene Symbol:
          Tmem60
          Gene Name:
          transmembrane protein 60
          Gene Aliases:
          BC027828
          Chromosome Location:
          Chr.5: 20882453 - 20886870 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617049_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem60
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177601.3 NP_808269.1 2 468 73
          GenBank mRNA BC027828.1 - 1 245 5249625 73

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem60

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23523161 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23523162 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23523160 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P404461 NM_177601.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P404461 AK041036.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P404461 BC027828.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P404461 NM_177601.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P404461 AK041036.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P404461 BC027828.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 696106 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 74734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431824 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154936 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P404461 NM_177601.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P404461 AK041036.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P404461 BC027828.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P404461 NM_177601.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P404461 AK041036.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P404461 BC027828.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23523194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23523187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23523193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23523186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23523189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23523196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23523197 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23523191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23523192 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154936 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05498706 - -
          Whole Mouse Genome G4122A A_52_P404461 NM_177601.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404461 AK041036.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404461 BC027828.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404461 NM_177601.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404461 AK041036.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P404461 BC027828.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104551 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04197013 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04504176 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04270942 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04270947 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04254938 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04504177 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04254943 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04270941 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23523160 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23523162 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23523161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23523180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04725242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04254926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523207 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523167 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04270936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04254946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523191 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04687264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04254936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523193 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523202 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523187 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04254935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523160 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23523180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04197009 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10519946 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10519948 - -
          Affymetrix HT MG-430 PM Array Plate 1458106_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428000_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10519945 - -
          Mouse Genome 430 2.0 Array 1428000_PM_at - -
          Mouse Genome 430 2.0 Array 1428000_at - -
          Mouse Genome 430 2.0 Array 1458106_PM_at - -
          Mouse Genome 430 2.0 Array 1458106_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 212090_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 212090_at - -
          Mouse Expression Set 430 1428000_PM_at - -
          Mouse Expression Set 430 1428000_at - -
          Mouse Expression Set 430 1458106_PM_at - -
          Mouse Expression Set 430 1458106_at - -
          Mouse Genome 430A 2.0 Array 1428000_PM_at - -
          Mouse Genome 430A 2.0 Array 1428000_at - -
          Affymetrix HT Mouse Genome 430A Array 1428000_at - -
          Affymetrix HT Mouse Genome 430A Array 1428000_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458106_at - -
          Affymetrix HT Mouse Genome 430B Array 1458106_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 626898 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106350129 - -
          Illumina mouse-6 v1.0 expression beadchip 7050600 - -
          Illumina mouse-6 v1.0 expression beadchip 106350129 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007050600 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620627 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2572759 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620627 - -
          mouse-6 v1.1 (Illumina) scl0212090.1_216-S - -
          Illumina Sentrix mouse-6.v1 bead array 7050600 - -
          Illumina Sentrix mouse-6.v1 bead array 106350129 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0212090.1_216-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050600 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106350129 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2813390 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2813390 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216762 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline