Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617112_m1
          See other PSTK GE Assays ›
          Gene Symbol:
          Pstk
          Gene Name:
          phosphoseryl-tRNA kinase
          Gene Aliases:
          5430423O14Rik, 5730458D16Rik, Gm1099, R75284
          Chromosome Location:
          Chr.7: 131371113 - 131392768 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617112_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pstk
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039534.1 NP_001034623.1 1 - 2 245 71
          GenBank mRNA AK077585.1 - 1 - 2 246 71
          BC110381.1 - 1 - 2 221 5149569 71
          BC115503.2 - 1 - 2 246 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001919.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          selenocysteine incorporation
          translation
          phosphorylation
          tRNA binding
          nucleotide binding
          ATP binding
          kinase activity
          transferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25528061 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25528063 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25528062 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487308 BC030313.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487308 BC110381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487308 BC115503.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487308 AK077585.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487308 NM_001039534.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487308 BC030313.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487308 BC110381.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487308 BC115503.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487308 AK077585.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P487308 NM_001039534.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105535 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 768012 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 341638 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 660306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 565539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 870335 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34442 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139133 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135647 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487308 BC030313.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487308 BC110381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487308 BC115503.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487308 AK077585.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487308 NM_001039534.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487308 BC030313.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487308 BC110381.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487308 BC115503.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487308 AK077585.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P487308 NM_001039534.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25528060 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135647 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139133 - -
          Whole Mouse Genome G4122A A_51_P355243 AK028520.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P355243 AK140401.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487308 BC030313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487308 BC110381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487308 BC115503.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487308 AK077585.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487308 NM_001039534.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487308 BC030313.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487308 BC110381.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487308 BC115503.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487308 AK077585.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P487308 NM_001039534.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P410529 AK017332.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135647 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139133 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06331085 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25528062 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25528063 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25528061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25527987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25528100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06201487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06487964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06628588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06603451 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06331083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05940705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528061 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06603450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06471936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06558921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06201485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06471945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25528039 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10558225 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558221 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558226 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558222 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558224 - -
          Affymetrix Mouse Gene 1.0 ST Array 10558223 - -
          Affymetrix HT MG-430 PM Array Plate 1434516_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455953_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10558220 - -
          Mouse Genome 430 2.0 Array 1434516_PM_at - -
          Mouse Genome 430 2.0 Array 1455953_PM_x_at - -
          Mouse Genome 430 2.0 Array 1434516_at - -
          Mouse Genome 430 2.0 Array 1455953_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214580_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 214580_at - -
          Affymetrix Mouse Clariom S Array PSR0700016839.mm.1 TC0700001919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700016839.mm.1 NM_001039534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016839.mm.1 NM_001039534.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016841.mm.1 TC0700001919.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700016841.mm.1 NM_001039534.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700016841.mm.1 NM_001039534.1 Refseq
          Mouse Expression Set 430 1434516_PM_at - -
          Mouse Expression Set 430 1434516_at - -
          Mouse Expression Set 430 1455953_x_at - -
          Mouse Expression Set 430 1455953_PM_x_at - -
          Murine Genome U74v2 Set 99190_PM_at - -
          Murine Genome U74v2 Set 99190_at - -
          Mouse Genome 430A 2.0 Array 1455953_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1455953_x_at - -
          Mouse Genome 430A 2.0 Array 1434516_at - -
          Mouse Genome 430A 2.0 Array 1434516_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455953_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1434516_at - -
          Affymetrix HT Mouse Genome 430A Array 1434516_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455953_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 919310 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2591908 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2591908 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline