Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617276_s1
          See other LDOC1L GE Assays ›
          Gene Symbol:
          Ldoc1l
          Gene Name:
          leucine zipper, down-regulated in cancer 1-like
          Gene Aliases:
          BC058638, Mar6, Mart6
          Chromosome Location:
          Chr.15: 84553398 - 84557823 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617276_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ldoc1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177630.3 NP_808298.2 1 3389 63
          GenBank mRNA AK083881.1 - 1 3351 63
          BC035491.1 - 1 2445 63
          BC058638.1 - 1 3058 6418256 63
          BC094559.1 - 1 3389 6826763 63

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ldoc1l

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30395526 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30395522 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30395524 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30395523 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011245 BC035491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011245 BC058638.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011245 BC094559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011245 NM_177630.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2011245 AK083881.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139615 BC035491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139615 BC058638.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139615 BC094559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139615 NM_177630.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2139615 AK083881.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P130727 BC035491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P130727 BC058638.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P130727 BC094559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P130727 NM_177630.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P130727 AK083881.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P130727 BC035491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P130727 BC058638.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P130727 BC094559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P130727 NM_177630.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P130727 AK083881.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 658107 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P135686 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139615 BC035491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139615 BC058638.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139615 BC094559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139615 NM_177630.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2139615 AK083881.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011245 BC035491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011245 BC058638.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011245 BC094559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011245 NM_177630.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2011245 AK083881.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P130727 BC035491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P130727 BC058638.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P130727 BC094559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P130727 NM_177630.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P130727 AK083881.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P130727 BC035491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P130727 BC058638.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P130727 BC094559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P130727 NM_177630.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P130727 AK083881.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395534 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395520 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395546 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395504 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395500 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395530 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395526 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395532 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395523 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30395549 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P135686 - -
          Whole Mouse Genome G4122A A_52_P130727 BC035491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P130727 BC058638.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P130727 BC094559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P130727 NM_177630.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P130727 AK083881.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P130727 BC035491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P130727 BC058638.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P130727 BC094559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P130727 NM_177630.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P130727 AK083881.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105289 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P135686 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12162333 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12162309 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12156962 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12162307 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12162291 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12162283 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12162353 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12156957 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12162322 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12162313 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12162340 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30395522 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30395526 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30395524 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30395523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395530 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30395461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395538 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12162301 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395505 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12162352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395473 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12103899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12162291 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12238167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12162308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12162302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395556 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395500 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395463 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11939435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395475 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395530 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11939437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12162336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395492 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395501 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395527 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30395487 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10431153 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431152 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10431149 - -
          Affymetrix HT MG-430 PM Array Plate 1423673_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10431147 - -
          Mouse Genome 430 2.0 Array 1423673_PM_at - -
          Mouse Genome 430 2.0 Array 1423673_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223732_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 223732_PM_at - -
          Mouse Expression Set 430 1423673_at - -
          Mouse Expression Set 430 1423673_PM_at - -
          Murine Genome U74v2 Set 93423_PM_at - -
          Murine Genome U74v2 Set 93423_at - -
          Mouse Genome 430A 2.0 Array 1423673_at - -
          Mouse Genome 430A 2.0 Array 1423673_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423673_at - -
          Affymetrix HT Mouse Genome 430A Array 1423673_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 422333 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6020086 - -
          Illumina mouse-6 v1.0 expression beadchip 6020086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006020086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2596499 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2596499 - -
          mouse-6 v1.1 (Illumina) scl46885.1.976_201-S - -
          Illumina Sentrix mouse-6.v1 bead array 6020086 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46885.1.976_201-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6020086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2596499 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2596499 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl46885.1.976_201-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline