Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617341_g1
          See other H2-M1 GE Assays ›
          Gene Symbol:
          H2-M1
          Gene Name:
          histocompatibility 2, M region locus 1
          Gene Aliases:
          H-2M1, H2-Qa-Mb1, Mb1
          Chromosome Location:
          Chr.17: 36669030 - 36672197 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617341_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: H2-M1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177636.2 NP_808304.2 4 - 5 902 105
          GenBank mRNA AF539594.1 - 4 - 5 902 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002031.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene H2-M1

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          antigen processing and presentation of peptide antigen via MHC class I
          receptor binding
          peptide antigen binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166177 NM_177636.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166177 AF539594.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 502452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947196 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108462 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174524 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155848 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166177 NM_177636.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166177 AF539594.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P181698 NM_177636.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P181698 AF539594.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31165078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31165063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31165071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31165079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31165082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31165067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31165054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31165064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31165084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31165059 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108462 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155848 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174524 - -
          Whole Mouse Genome G4122A A_52_P181698 NM_177636.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P181698 AF539594.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124207 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155848 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174524 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108462 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13546588 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13546561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13546585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31165067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165063 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165082 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13530703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13546561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13082948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165090 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165059 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165083 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165050 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165048 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31165085 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450791 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450793 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450795 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450792 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450794 - -
          Affymetrix Mouse Gene 1.1 ST Array 10450790 - -
          Affymetrix Mouse Clariom S Array PSR1700019399.mm.1 NM_177636.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019399.mm.1 NM_177636.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019399.mm.1 TC1700002031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019398.mm.1 NM_177636.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019398.mm.1 NM_177636.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019398.mm.1 TC1700002031.mm.1 MTA TC
          Murine Genome U74v2 Set 101299_at - -
          Murine Genome U74v2 Set 101299_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 873989 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3440739 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003440739 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2659185 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2659185 - -
          mouse-6 v1.1 (Illumina) scl49949.5.1_15-S - -
          Illumina Sentrix mouse-6.v1 bead array 3440739 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49949.5.1_15-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3440739 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2659185 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2659185 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0