Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617456_m1
          See other R3HDM4 GE Assays ›
          Gene Symbol:
          R3hdm4
          Gene Name:
          R3H domain containing 4
          Gene Aliases:
          AI503502, C030046I01Rik
          Chromosome Location:
          Chr.10: 79910052 - 79916941 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617456_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: R3hdm4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177994.4 NP_818775.2 7 - 8 735 74
          GenBank mRNA AK081288.1 - 7 - 8 724 74
          AK088107.1 - 7 - 8 736 74
          AK139047.1 - 7 - 8 726 74
          AK159869.1 - 7 - 8 713 74
          AK165610.1 - 7 - 8 706 74
          BC034653.1 - 6 - 7 538 74
          BC095970.1 - 7 - 8 716 6530790 74
          BC132463.1 - 7 - 8 811 40130740 74
          BC138461.1 - 7 - 8 811 8861481 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002485.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene R3hdm4

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          nucleic acid binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27279700 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P623058 BC095970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P623058 BC132463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P623058 BC138461.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P623058 BC034653.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P623058 BC040686.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P623058 BC095970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P623058 BC132463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P623058 BC138461.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P623058 BC034653.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P623058 BC040686.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652362 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 335142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622001 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37833 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152133 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P623058 BC095970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P623058 BC132463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P623058 BC138461.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P623058 BC034653.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P623058 BC040686.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P623058 BC095970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P623058 BC132463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P623058 BC138461.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P623058 BC034653.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P623058 BC040686.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279689 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27279655 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152133 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07120849 - -
          Whole Mouse Genome G4122A A_52_P623058 BC095970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P623058 BC132463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P623058 BC138461.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P623058 BC034653.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P623058 BC040686.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P623058 BC095970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P623058 BC132463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P623058 BC138461.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P623058 BC034653.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P623058 BC040686.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117630 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P130862 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152133 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08369225 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08369227 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27279700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27279691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08521456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279653 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08369228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279669 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279670 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08369224 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279655 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08521450 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08369226 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08521448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08418236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08418221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27279681 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10370682 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370684 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370683 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370689 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370685 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370687 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370686 - -
          Affymetrix Mouse Gene 1.0 ST Array 10370688 - -
          Affymetrix HT MG-430 PM Array Plate 1433931_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1456564_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433932_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1438905_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10370681 - -
          Mouse Genome 430 2.0 Array 1438905_x_at - -
          Mouse Genome 430 2.0 Array 1456564_PM_at - -
          Mouse Genome 430 2.0 Array 1433932_PM_x_at - -
          Mouse Genome 430 2.0 Array 1438905_PM_x_at - -
          Mouse Genome 430 2.0 Array 1456564_at - -
          Mouse Genome 430 2.0 Array 1433931_at - -
          Mouse Genome 430 2.0 Array 1433931_PM_at - -
          Mouse Genome 430 2.0 Array 1433932_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 109284_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 109284_at - -
          Affymetrix Mouse Clariom S Array PSR1000018234.mm.1 NM_177994.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018234.mm.1 NM_177994.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018234.mm.1 TC1000002485.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018238.mm.1 NM_177994.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018238.mm.1 NM_177994.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018238.mm.1 TC1000002485.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018235.mm.1 NM_177994.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018235.mm.1 NM_177994.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018235.mm.1 TC1000002485.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018236.mm.1 NM_177994.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018236.mm.1 NM_177994.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018236.mm.1 TC1000002485.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018233.mm.1 NM_177994.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018233.mm.1 NM_177994.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018233.mm.1 TC1000002485.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000018237.mm.1 NM_177994.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018237.mm.1 NM_177994.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000018237.mm.1 TC1000002485.mm.1 MTA TC
          Mouse Expression Set 430 1438905_x_at - -
          Mouse Expression Set 430 1456564_PM_at - -
          Mouse Expression Set 430 1438905_PM_x_at - -
          Mouse Expression Set 430 1456564_at - -
          Mouse Expression Set 430 1433931_PM_at - -
          Mouse Expression Set 430 1433932_PM_x_at - -
          Mouse Expression Set 430 1433931_at - -
          Mouse Expression Set 430 1433932_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1433931_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433932_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1456564_at - -
          Affymetrix HT Mouse Genome 430B Array 1433931_at - -
          Affymetrix HT Mouse Genome 430B Array 1438905_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1438905_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1433932_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1456564_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3120292 - -
          Illumina mouse-6 v1.0 expression beadchip 3120292 - -
          Illumina mouse-6 v1.0 expression beadchip 102480100 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003120292 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224551 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244149 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224551 - -
          mouse-6 v1.1 (Illumina) scl00109284.2_77-S - -
          mouse-6 v1.1 (Illumina) scl37746.9_45-S - -
          Illumina Sentrix mouse-6.v1 bead array 3120292 - -
          Illumina Sentrix mouse-6.v1 bead array 102480100 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00109284.2_77-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3120292 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102480100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1224551 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1244149 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224551 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00109284.2_77-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline