Search
Search
| Gene Symbol: Etl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081006.1 | NP_001074475.1 | 9 - 10 | 2077 | 60 | |
| NM_001177630.2 | NP_001171101.2 | 8 - 9 | 2175 | 60 | ||
| NM_001177631.1 | NP_001171102.1 | 7 - 8 | 2070 | 60 | ||
| NM_029895.4 | NP_084171.2 | 8 - 9 | 2283 | 60 | ||
| NM_178059.5 | NP_835160.2 | 8 - 9 | 2283 | 60 | ||
| GenBank mRNA | AB093289.2 | - | 7 - 8 | 1860 | 60 | |
| AB125594.2 | - | 8 - 9 | 2283 | 60 | ||
| AB125595.2 | - | 8 - 9 | 2283 | 60 | ||
| AK054453.1 | - | 3 - 4 | 1226 | 60 | ||
| AK086078.1 | - | 3 - 4 | 1228 | 60 | ||
| AK147510.1 | - | 7 - 8 | 1977 | 60 | ||
| BC158051.1 | - | 8 - 9 | 1897 | 9007405 | 60 | |
| BC172143.1 | - | 7 - 8 | 1790 | 60 | ||
Set Membership: |
|