Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00617681_m1
          See other ZSWIM3 GE Assays ›
          Gene Symbol:
          Zswim3
          Gene Name:
          zinc finger SWIM-type containing 3
          Gene Aliases:
          4921517A06Rik, C86566
          Chromosome Location:
          Chr.2: 164805114 - 164822127 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00617681_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zswim3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178375.2 NP_848462.1 1 - 2 318 70
          GenBank mRNA AK014904.1 - 1 - 2 338 70
          BC023287.1 - 1 - 2 318 3993352 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002534.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zswim3

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P59228 BC023287.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P59228 NM_178375.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P59228 AK014904.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P59228 BC023287.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P59228 NM_178375.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P59228 AK014904.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 447392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104830 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943445 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 402534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118618 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 827551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387834 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 639887 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230483 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82105 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P116467 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167211 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P59228 BC023287.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P59228 NM_178375.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P59228 AK014904.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P59228 BC023287.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P59228 NM_178375.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P59228 AK014904.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819934 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819879 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P116467 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00550471 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00550468 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167211 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04773057 - -
          Whole Mouse Genome G4122A A_52_P59228 BC023287.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P59228 NM_178375.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P59228 AK014904.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P59228 BC023287.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P59228 NM_178375.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P59228 AK014904.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104022 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P116467 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167211 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02070581 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01261419 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01261418 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02070587 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02070592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02070592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819917 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01313270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01865072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01865071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819920 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02070590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01268894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819879 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02070587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01261441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01979565 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819933 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819867 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01941014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01251058 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01261428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819934 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01979566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819937 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819899 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819901 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02173176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01261434 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10478589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478588 - -
          Affymetrix HT MG-430 PM Array Plate 1428313_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10478587 - -
          Mouse Genome 430 2.0 Array 1457785_at - -
          Mouse Genome 430 2.0 Array 1457785_PM_at - -
          Mouse Genome 430 2.0 Array 1444571_PM_at - -
          Mouse Genome 430 2.0 Array 1444571_at - -
          Mouse Genome 430 2.0 Array 1428313_PM_at - -
          Mouse Genome 430 2.0 Array 1428313_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67538_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67538_at - -
          Affymetrix Mouse Clariom S Array PSR0200020723.mm.1 NM_178375.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020723.mm.1 NM_178375.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020723.mm.1 TC0200002534.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020722.mm.1 NM_178375.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020722.mm.1 NM_178375.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020722.mm.1 TC0200002534.mm.1 MTA TC
          Mouse Expression Set 430 1428313_at - -
          Mouse Expression Set 430 1428313_PM_at - -
          Murine Genome U74v2 Set 165679_PM_at - -
          Murine Genome U74v2 Set 165679_at - -
          Affymetrix HT Mouse Genome 430B Array 1428313_at - -
          Affymetrix HT Mouse Genome 430B Array 1428313_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 472351 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1230563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001230563 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252526 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252526 - -
          mouse-6 v1.1 (Illumina) scl19922.2_573-S - -
          Illumina Sentrix mouse-6.v1 bead array 1230563 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19922.2_573-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1230563 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1252526 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252526 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-6wvps:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0