Search
Search

Fam53a| Gene Symbol: Fam53a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178390.3 | NP_848477.2 | 3 - 4 | 659 | 55 | |
| GenBank mRNA | AF396665.1 | - | 1 - 2 | 81 | 55 | |
| AK010537.1 | - | 3 - 4 | 596 | 55 | ||
| AK017321.1 | - | 2 - 3 | 1002 | 55 | ||
| AK086386.1 | - | 3 - 4 | 624 | 55 | ||
| AK155077.1 | - | 3 - 4 | 613 | 55 | ||
| AK163326.1 | - | 3 - 4 | 630 | 55 | ||
| AK220276.1 | - | 2 - 3 | 295 | 55 | ||
| BC016596.1 | - | 1 - 2 | 84 | 3257498 | 55 | |
| BC028656.1 | - | 3 - 4 | 379 | 1244982 | 55 | |
Set Membership: |
|