Search
Search

Atp1a2| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178405.3 | NP_848492.1 | 20 - 21 | 3012 | 69 | |
| GenBank mRNA | AK129212.1 | - | 20 - 21 | 2925 | 69 | |
| AK137602.1 | - | 20 - 21 | 2948 | 69 | ||
| AK147331.1 | - | 20 - 21 | 2944 | 69 | ||
| AK148775.1 | - | 19 - 20 | 3134 | 69 | ||
| AK161840.1 | - | 20 - 21 | 2944 | 69 | ||
| AK164509.1 | - | 20 - 21 | 2948 | 69 | ||
| AK168703.1 | - | 20 - 21 | 2944 | 69 | ||
| BC025807.1 | - | 9 - 10 | 1240 | 69 | ||
| BC036127.1 | - | 20 - 21 | 2936 | 4955003 | 69 | |
| BC041774.1 | - | 17 - 18 | 2662 | 69 | ||
Set Membership: |
|