Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00618072_g1
          See other NOP16 GE Assays ›
          Gene Symbol:
          Nop16
          Gene Name:
          NOP16 nucleolar protein
          Gene Aliases:
          AA409471, D13Wsu177e
          Chromosome Location:
          Chr.13: 54584191 - 54590074 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00618072_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nop16
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178605.4 NP_848720.1 1 - 2 212 73
          GenBank mRNA AK002696.1 - 1 - 2 160 73
          AK013251.1 - 1 - 2 213 73
          AK088248.1 - 1 - 2 213 73
          BC012213.1 - 1 - 2 188 3964770 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002036.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nop16

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          ribosomal large subunit biogenesis
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29050255 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29050252 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29050253 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P622434 AK003769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P622434 AK013251.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P622434 AK088248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P622434 BC012213.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P622434 NM_178605.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P622434 AK003769.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P622434 AK013251.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P622434 AK088248.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P622434 BC012213.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P622434 NM_178605.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360988 AK033092.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360988 AK045093.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360988 AK135662.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360988 AK155895.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360988 AK160163.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360988 BC086794.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 360988 NM_008974.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799450 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133839 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P622434 AK003769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P622434 AK013251.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P622434 AK088248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P622434 BC012213.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P622434 NM_178605.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P622434 AK003769.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P622434 AK013251.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P622434 AK088248.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P622434 BC012213.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P622434 NM_178605.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29050246 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133839 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02695455 - -
          Whole Mouse Genome G4122A A_52_P622434 AK003769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P622434 AK013251.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P622434 AK088248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P622434 BC012213.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P622434 NM_178605.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P622434 AK003769.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P622434 AK013251.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P622434 AK088248.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P622434 BC012213.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P622434 NM_178605.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P474158 AK013251.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P474158 NM_178605.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117746 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133839 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10440107 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10854346 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10854351 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10854348 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29050253 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29050255 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29050252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29050146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10854342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10705757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11071366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050247 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10705755 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10705753 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10680451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050254 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10936089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10716186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29050215 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409340 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409339 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409344 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409341 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409342 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409343 - -
          Affymetrix HT MG-430 PM Array Plate 1451414_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1424620_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409338 - -
          Mouse Genome 430 2.0 Array 1451414_PM_at - -
          Mouse Genome 430 2.0 Array 1451414_at - -
          Mouse Genome 430 2.0 Array 1424620_PM_at - -
          Mouse Genome 430 2.0 Array 1424620_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 28126_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 28126_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300012722.mm.1 NM_178605.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012722.mm.1 NM_178605.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012722.mm.1 TC1300002036.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012718.mm.1 NM_178605.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012718.mm.1 NM_178605.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012718.mm.1 TC1300002036.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012719.mm.1 NM_178605.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012719.mm.1 NM_178605.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012719.mm.1 TC1300002036.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012721.mm.1 NM_178605.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012721.mm.1 NM_178605.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012721.mm.1 TC1300002036.mm.1 MTA TC
          Mouse Expression Set 430 1424620_PM_at - -
          Mouse Expression Set 430 1451414_at - -
          Mouse Expression Set 430 1424620_at - -
          Mouse Expression Set 430 1451414_PM_at - -
          Murine Genome U74v2 Set 95657_f_at - -
          Murine Genome U74v2 Set 95657_PM_f_at - -
          Murine Genome U74v2 Set 95656_PM_i_at - -
          Murine Genome U74v2 Set 95656_i_at - -
          Murine Genome U74v2 Set 168567_at - -
          Murine Genome U74v2 Set 168567_PM_at - -
          Mouse Genome 430A 2.0 Array 1424620_at - -
          Mouse Genome 430A 2.0 Array 1451414_PM_at - -
          Mouse Genome 430A 2.0 Array 1424620_PM_at - -
          Mouse Genome 430A 2.0 Array 1451414_at - -
          Affymetrix HT Mouse Genome 430A Array 1451414_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424620_at - -
          Affymetrix HT Mouse Genome 430A Array 1451414_at - -
          Affymetrix HT Mouse Genome 430A Array 1424620_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 656624 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1850215 - -
          Illumina mouse-6 v1.0 expression beadchip 1850215 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001850215 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227079 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227079 - -
          mouse-6 v1.1 (Illumina) scl43939.4_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 1850215 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43939.4_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1850215 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1227079 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227079 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline