Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00618116_m1
          See other CNPY4 GE Assays ›
          Gene Symbol:
          Cnpy4
          Gene Name:
          canopy 4 homolog (zebrafish)
          Gene Aliases:
          2610019P18Rik
          Chromosome Location:
          Chr.5: 138187535 - 138193894 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00618116_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cnpy4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178612.4 NP_848727.1 3 - 4 404 65
          GenBank mRNA AK011474.1 - 3 - 4 405 65
          AK012968.1 - 3 - 4 386 65
          AK033219.1 - 3 - 4 406 65
          AK051562.1 - 3 - 4 406 65
          BC086647.1 - 3 - 4 369 6818372 65
          BC094672.1 - 3 - 4 511 30633103 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001645.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cnpy4

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of protein localization
          receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161400 BC094672.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161400 AK033219.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161400 AK051562.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161400 AK012968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161400 NM_178612.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161400 BC094672.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161400 AK033219.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161400 AK051562.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161400 AK012968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P161400 NM_178612.4 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806072 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582132 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 972797 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20001 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160907 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161400 BC094672.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161400 AK033219.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161400 AK051562.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161400 AK012968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161400 NM_178612.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161400 BC094672.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161400 AK033219.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161400 AK051562.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161400 AK012968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P161400 NM_178612.4 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129275 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24129303 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01232728 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160907 - -
          Whole Mouse Genome G4122A A_52_P161400 BC094672.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161400 AK033219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161400 AK051562.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161400 AK012968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161400 NM_178612.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161400 BC094672.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161400 AK033219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161400 AK051562.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161400 AK012968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P161400 NM_178612.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P243194 AK011474.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127279 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160907 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04596099 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04596100 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04189582 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04189584 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04596096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24129273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04648702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04927626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04497677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129275 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04189582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04189584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04699619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04596096 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04648704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04775975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04699622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129293 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04596100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24129298 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10526777 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526773 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526779 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526778 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526776 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526780 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526774 - -
          Affymetrix Mouse Gene 1.0 ST Array 10526775 - -
          Affymetrix HT MG-430 PM Array Plate 1428325_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10526772 - -
          Mouse Genome 430 2.0 Array 1428325_PM_at - -
          Mouse Genome 430 2.0 Array 1428325_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66455_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66455_at - -
          Affymetrix Mouse Clariom S Array PSR0500015102.mm.1 TC0500001645.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015102.mm.1 NM_178612.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015102.mm.1 NM_178612.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015104.mm.1 TC0500001645.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015104.mm.1 NM_178612.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015104.mm.1 NM_178612.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015100.mm.1 TC0500001645.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015100.mm.1 NM_178612.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015100.mm.1 NM_178612.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015105.mm.1 TC0500001645.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015105.mm.1 NM_178612.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015105.mm.1 NM_178612.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015103.mm.1 TC0500001645.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500015103.mm.1 NM_178612.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500015103.mm.1 NM_178612.4 Refseq
          Mouse Expression Set 430 1428325_at - -
          Mouse Expression Set 430 1428325_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428325_at - -
          Affymetrix HT Mouse Genome 430B Array 1428325_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 842560 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4570711 - -
          Illumina mouse-6 v1.0 expression beadchip 4570711 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004570711 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2673052 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2673052 - -
          mouse-6 v1.1 (Illumina) scl27081.7_488-S - -
          Illumina Sentrix mouse-6.v1 bead array 4570711 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl27081.7_488-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4570711 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2843403 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2843403 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline