Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00618358_m1
          See other RBMS3 GE Assays ›
          Gene Symbol:
          Rbms3
          Gene Name:
          RNA binding motif, single stranded interacting protein
          Gene Aliases:
          6720477E09Rik, 8430436O14Rik
          Chromosome Location:
          Chr.9: 116572746 - 118040292 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00618358_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rbms3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001172121.1 NP_001165592.1 8 - 9 944 121
          NM_001172122.1 NP_001165593.1 7 - 8 1416 121
          NM_001172123.1 NP_001165594.1 7 - 8 1416 121
          NM_001172124.1 NP_001165595.1 7 - 8 1416 121
          NM_001172126.1 NP_001165597.1 7 - 8 1416 121
          NM_178660.4 NP_848775.2 7 - 8 1416 121
          GenBank mRNA AK030478.1 - 7 - 8 1076 121
          AK030609.1 - 7 - 8 1050 121
          AK032938.1 - 7 - 8 1413 121
          AK033005.1 - 7 - 8 1059 121
          AK041098.1 - 7 - 8 1418 121
          AK046197.1 - 8 - 9 945 121
          AK051776.1 - 8 - 9 1463 121
          AK052196.1 - 7 - 8 1397 121
          BC033604.1 - 3 - 4 271 121
          BC108402.1 - 7 - 8 1390 6516420 121
          BC117841.1 - 7 - 8 856 40093225 121

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003204.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rbms3

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of translation
          nucleotide binding
          nucleic acid binding
          RNA binding
          mRNA 3'-UTR binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_001172121.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_001172121.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_001172122.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_001172122.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_001172123.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_001172123.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_001172124.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_001172124.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_001172126.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_001172126.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_178660.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 NM_178660.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900027026.mm.1 TC0900003204.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline