Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00618407_m1
          See other TFDP2 GE Assays ›
          Gene Symbol:
          Tfdp2
          Gene Name:
          transcription factor Dp 2
          Gene Aliases:
          1110029I05Rik, A330080J22Rik, DP-3, DP3
          Chromosome Location:
          Chr.9: 96196275 - 96323646 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00618407_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tfdp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001184706.1 NP_001171635.1 8 - 9 1031 78
          NM_001184708.1 NP_001171637.1 5 - 6 757 78
          NM_001184709.1 NP_001171638.1 6 - 7 655 78
          NM_001184710.1 NP_001171639.1 5 - 6 531 78
          NM_001184711.1 NP_001171640.1 6 - 7 638 78
          NM_178667.4 NP_848782.2 9 - 10 1032 78
          GenBank mRNA AK029055.1 - 8 - 9 1004 78
          AK033356.1 - 6 - 7 619 78
          AK044784.1 - 5 - 6 759 78
          AK053881.1 - 8 - 9 990 78
          AK079624.1 - 9 - 10 960 78
          AK158829.1 - 5 - 6 527 78
          BC092230.1 - 6 - 7 601 78
          BC141080.1 - 7 - 8 689 9055692 78
          CA980067.1 - 6 - 7 638 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001210.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tfdp2

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cell cycle
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          cell cycle
          negative regulation of transcription, DNA-templated
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription cofactor activity
          protein binding
          transcription factor binding
          protein domain specific binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900009540.mm.1 NM_001184709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009540.mm.1 NM_001184709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009540.mm.1 TC0900001210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_001184706.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_001184706.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_001184708.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_001184708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_001184709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_001184709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_001184710.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_001184710.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_001184711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_001184711.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_178667.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 NM_178667.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009550.mm.1 TC0900001210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_001184706.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_001184706.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_001184708.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_001184708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_001184709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_001184709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_001184710.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_001184710.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_001184711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_001184711.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_178667.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 NM_178667.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009551.mm.1 TC0900001210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900009541.mm.1 NM_001184710.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009541.mm.1 NM_001184710.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009541.mm.1 NM_001184711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009541.mm.1 NM_001184711.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009541.mm.1 TC0900001210.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_001184706.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_001184706.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_001184708.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_001184708.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_001184709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_001184709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_001184710.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_001184710.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_001184711.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_001184711.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_178667.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 NM_178667.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900009553.mm.1 TC0900001210.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 834714 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline