| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10430930 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1456671_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10430929 |
- |
- |
| Mouse Genome 430 2.0 Array |
1456671_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1456671_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1500015431.mm.1 |
NR_027799.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015431.mm.1 |
NR_027799.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015431.mm.1 |
TC1500001960.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015430.mm.1 |
NR_027799.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015430.mm.1 |
NR_027799.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015430.mm.1 |
TC1500001960.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015429.mm.1 |
NR_027799.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015429.mm.1 |
NR_027799.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015429.mm.1 |
TC1500001960.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1500015432.mm.1 |
NR_027799.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015432.mm.1 |
NR_027799.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1500015432.mm.1 |
TC1500001960.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1456671_PM_at |
- |
- |
| Mouse Expression Set 430 |
1456671_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1456671_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1456671_PM_at |
- |
- |