Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00620237_m1
          See other DPCD GE Assays ›
          Gene Symbol:
          Dpcd
          Gene Name:
          deleted in primary ciliary dyskinesia
          Gene Aliases:
          5330431N19Rik, Ndac
          Chromosome Location:
          Chr.19: 45560590 - 45578287 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00620237_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dpcd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_172639.2 NP_766227.1 2 - 3 170 86
          GenBank mRNA AK077347.1 - 2 - 3 171 86
          BC027559.1 - 2 - 3 182 1493971 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000675.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dpcd

          Gene Ontology Categories:

          Function(s) Process(es)

          epithelial cilium movement
          spermatogenesis
          determination of left/right symmetry
          ventricular system development
          lateral ventricle development
          third ventricle development
          sperm motility
          left/right pattern formation
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P37498 NM_172639.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P37498 AK077347.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P37498 BC027559.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P37498 NM_172639.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P37498 AK077347.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P37498 BC027559.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580493 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 505810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 24054 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858724 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 929745 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345174 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961526 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63506 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171736 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166620 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P37498 NM_172639.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P37498 AK077347.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P37498 BC027559.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P37498 NM_172639.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P37498 AK077347.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P37498 BC027559.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139184 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32139165 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03605383 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166620 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171736 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03605376 - -
          Whole Mouse Genome G4122A A_52_P37498 NM_172639.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37498 AK077347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37498 BC027559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37498 NM_172639.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37498 AK077347.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P37498 BC027559.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369294 NM_172639.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369294 AK077347.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P118283 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166620 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171736 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14300077 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14168955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139275 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32139221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14218628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14481338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14481356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14144108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14246862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14354523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139274 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14330218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139262 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139218 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139205 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139258 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14481343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139166 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14380513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139272 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14330216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14481324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14481362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14246864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139263 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14235465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14150683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14481379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14208895 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14481323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139249 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32139248 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10463508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463507 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10463506 - -
          Affymetrix HT MG-430 PM Array Plate 1452603_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10463505 - -
          Mouse Genome 430 2.0 Array 1452603_at - -
          Mouse Genome 430 2.0 Array 1442674_at - -
          Mouse Genome 430 2.0 Array 1452603_PM_at - -
          Mouse Genome 430 2.0 Array 1442674_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226162_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 226162_at - -
          Affymetrix Mouse Clariom S Array PSR1900005951.mm.1 NM_172639.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005951.mm.1 NM_172639.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900005951.mm.1 TC1900000675.mm.1 MTA TC
          Mouse Expression Set 430 1452603_at - -
          Mouse Expression Set 430 1452603_PM_at - -
          Murine Genome U74v2 Set 104120_at - -
          Murine Genome U74v2 Set 104120_PM_at - -
          Mouse Genome 430A 2.0 Array 1452603_at - -
          Mouse Genome 430A 2.0 Array 1452603_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452603_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452603_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 576732 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2120452 - -
          Illumina mouse-6 v1.0 expression beadchip 2120452 - -
          Illumina mouse-6 v1.0 expression beadchip 780632 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000780632 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002120452 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2705644 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237049 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237049 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2705644 - -
          mouse-6 v1.1 (Illumina) scl0226162.6_83-S - -
          mouse-6 v1.1 (Illumina) scl000481.1_29-S - -
          Illumina Sentrix mouse-6.v1 bead array 780632 - -
          Illumina Sentrix mouse-6.v1 bead array 2120452 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000481.1_29-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0226162.6_83-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 780632 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2120452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2892480 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2892479 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237049 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237049 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2892479 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2892480 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000481.1_29-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline