Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00620413_m1
          See other SAMD11 GE Assays ›
          Gene Symbol:
          Samd11
          Gene Name:
          sterile alpha motif domain containing 11
          Gene Aliases:
          BC026991, mr-s
          Chromosome Location:
          Chr.4: 156246966 - 156255657 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00620413_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Samd11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001110516.1 NP_001103986.1 4 - 5 511 90
          GenBank mRNA AK149268.1 - 4 - 5 511 90
          AK161035.1 - 3 - 4 525 90
          AY458844.1 - 3 - 4 328 90
          BC026511.1 - 1 - 2 166 90
          BC026991.1 - 4 - 5 482 5363470 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004175.mm.1

          Set Membership:

          Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          chromatin/chromatin-binding, or -regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          SAM domain binding
          PH domain binding
          protein self-association

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972277 NM_001110516.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972277 AK149268.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972277 NM_001110516.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972277 AK149268.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102393 BC026511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102393 BC026991.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102393 BC026511.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P102393 BC026991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P102393 BC026511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P102393 BC026991.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P102393 BC026511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P102393 BC026991.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1456977_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1425288_PM_at - -
          Mouse Genome 430 2.0 Array 1456977_PM_at - -
          Mouse Genome 430 2.0 Array 1425288_at - -
          Mouse Genome 430 2.0 Array 1425288_PM_at - -
          Mouse Genome 430 2.0 Array 1456977_at - -
          Affymetrix Mouse Clariom S Array PSR0400035153.mm.1 TC0400004175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035153.mm.1 NM_001110516.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035153.mm.1 NM_001110516.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035156.mm.1 TC0400004175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035156.mm.1 NM_001110516.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035156.mm.1 NM_001110516.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035146.mm.1 TC0400004175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035146.mm.1 NM_001110516.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035146.mm.1 NM_001110516.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035151.mm.1 TC0400004175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035151.mm.1 NM_001110516.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035151.mm.1 NM_001110516.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035147.mm.1 TC0400004175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035147.mm.1 NM_001110516.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035147.mm.1 NM_001110516.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035148.mm.1 TC0400004175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035148.mm.1 NM_001110516.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035148.mm.1 NM_001110516.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035149.mm.1 TC0400004175.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400035149.mm.1 NM_001110516.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400035149.mm.1 NM_001110516.1 Refseq
          Mouse Expression Set 430 1425288_PM_at - -
          Mouse Expression Set 430 1425288_at - -
          Mouse Expression Set 430 1456977_PM_at - -
          Mouse Expression Set 430 1456977_at - -
          Mouse Genome 430A 2.0 Array 1425288_at - -
          Mouse Genome 430A 2.0 Array 1425288_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1425288_at - -
          Affymetrix HT Mouse Genome 430A Array 1425288_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456977_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1456977_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 913450 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5130047 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216690 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216690 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1216690 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1216690 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline