Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00620597_m1
          See other NEU4 GE Assays ›
          Gene Symbol:
          Neu4
          Gene Name:
          sialidase 4
          Gene Aliases:
          9330166I04
          Chromosome Location:
          Chr.1: 94020446 - 94028334 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00620597_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Neu4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_173772.3 NP_776133.1 3 - 4 613 60
          GenBank mRNA AB510404.1 - 4 - 5 594 60
          AK034236.1 - 3 - 4 584 60
          AY258421.1 - 3 - 4 526 60
          BC070438.1 - 3 - 4 613 6842155 60

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000954.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Neu4

          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          glycoprotein catabolic process
          lipid metabolic process
          ganglioside catabolic process
          metabolic process
          oligosaccharide catabolic process
          lipid catabolic process
          exo-alpha-sialidase activity
          hydrolase activity
          hydrolase activity, acting on glycosyl bonds
          exo-alpha-(2->3)-sialidase activity
          exo-alpha-(2->6)-sialidase activity
          exo-alpha-(2->8)-sialidase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074326 BC070438.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074326 AK047397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074326 NM_173772.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281090 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742045 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 11021 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952722 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P134386 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074326 BC070438.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074326 AK047397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074326 NM_173772.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P507709 AY258421.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P507709 BC070438.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P507709 AB510404.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P507709 AK034236.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P507709 NM_173772.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457760 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457771 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20457784 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00136938 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P134386 - -
          Whole Mouse Genome G4122A A_51_P507709 AY258421.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P507709 BC070438.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P507709 AB510404.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P507709 AK034236.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P507709 NM_173772.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120365 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101383 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P134386 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00679057 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00679052 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00679044 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00679046 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00679053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20457795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00679057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00587618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00679053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00258724 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00679040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00587614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00679032 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00587617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00679036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457754 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01077859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00587616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00879916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457771 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457769 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00587626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00352630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00679052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00879918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00587621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00683243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457790 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00445985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00587620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00381921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20457809 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10348911 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348915 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348914 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348913 - -
          Affymetrix Mouse Gene 1.0 ST Array 10348916 - -
          Affymetrix Mouse Gene 1.1 ST Array 10348910 - -
          Affymetrix Mouse Clariom S Array PSR0100008120.mm.1 NM_173772.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008120.mm.1 NM_173772.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0100008120.mm.1 TC0100000954.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 838866 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 430593 - -
          MouseWG-6 v2 Expression BeadChip 510113 - -
          Illumina mouse-6 v1.0 expression beadchip 430593 - -
          Illumina mouse-6 v1.0 expression beadchip 510113 - -
          Illumina mouse-6 v1.0 expression beadchip 2450279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002450279 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000430593 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000510113 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2686167 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605810 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2716254 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605810 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2716254 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2686167 - -
          mouse-6 v1.1 (Illumina) scl0241159.4_2-S - -
          mouse-6 v1.1 (Illumina) scl17615.4.1_258-S - -
          mouse-6 v1.1 (Illumina) scl00241159.1_279-S - -
          Illumina Sentrix mouse-6.v1 bead array 2450279 - -
          Illumina Sentrix mouse-6.v1 bead array 430593 - -
          Illumina Sentrix mouse-6.v1 bead array 510113 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl17615.4.1_258-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0241159.4_2-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00241159.1_279-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 510113 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 430593 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2450279 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2686167 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605810 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2716254 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2686167 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2716254 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605810 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00241159.1_279-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl17615.4.1_258-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0241159.4_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline