| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.1 ST Array |
10551435 |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0700003949.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003949.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003949.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003950.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003950.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003950.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003952.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003952.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003952.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003936.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003936.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003936.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003948.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003948.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003948.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003951.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003951.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003951.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003943.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003943.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003943.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003947.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003947.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003947.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003945.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003945.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003945.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003941.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003941.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003941.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003944.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003944.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003944.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003946.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003946.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003946.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003939.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003939.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003939.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003940.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003940.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003940.mm.1 |
NM_001164655.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003942.mm.1 |
TC0700000486.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700003942.mm.1 |
NM_001164655.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700003942.mm.1 |
NM_001164655.1 |
Refseq |
| Mouse Expression Set 430 |
1426872_at |
- |
- |
| Mouse Expression Set 430 |
1426872_PM_at |
- |
- |
| Murine Genome U74v2 Set |
97826_PM_at |
- |
- |
| Murine Genome U74v2 Set |
162286_r_at |
- |
- |
| Murine Genome U74v2 Set |
97826_at |
- |
- |
| Murine Genome U74v2 Set |
162286_PM_r_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1426872_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1426872_at |
- |
- |