| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1400020249.mm.1 |
NM_001081252.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020249.mm.1 |
NM_001081252.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020249.mm.1 |
TC1400002723.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400020251.mm.1 |
NM_001081252.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020251.mm.1 |
NM_001081252.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020251.mm.1 |
TC1400002723.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400020250.mm.1 |
NM_001081252.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020250.mm.1 |
NM_001081252.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020250.mm.1 |
TC1400002723.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1400020248.mm.1 |
NM_001081252.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020248.mm.1 |
NM_001081252.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1400020248.mm.1 |
TC1400002723.mm.1 |
MTA TC |