| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800012115.mm.1 |
NR_045696.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012115.mm.1 |
NR_045696.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012115.mm.1 |
TC0800001488.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800012113.mm.1 |
NR_045696.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012113.mm.1 |
NR_045696.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012113.mm.1 |
TC0800001488.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800012114.mm.1 |
NR_045696.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012114.mm.1 |
NR_045696.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800012114.mm.1 |
TC0800001488.mm.1 |
MTA TC |