Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00621296_s1
          See other A430010J10RIK GE Assays ›
          Gene Symbol:
          A430010J10Rik
          Gene Name:
          RIKEN cDNA A430010J10 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.6: 67157801 - 67169010 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00621296_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: A430010J10Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK083337.1 - 1 754 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610289 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156124 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 90735 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 410529 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 717868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6024 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 451196 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 750372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 893752 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 620199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 395382 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834083 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 212950 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625067 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 589452 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 244772 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720810 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 53048 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 175442 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 785359 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126699 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 101180 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05928769 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05928866 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05928834 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05928856 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05928802 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01349552 - -
          Whole Mouse Genome G4122A A_52_P105458 AK039826.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P105458 AK083337.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05928802 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05263007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05697875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05133478 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05588424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05601554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05017902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520171 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05709175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05771950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05752511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05666467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519911 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05133485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520086 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05173576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05688192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05173582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05126858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05097519 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05133489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05449698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05133492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519961 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05133471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05597563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05743488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05133497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05792596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05752526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05103573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05666466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519909 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05103571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05727298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519905 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05792597 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05133467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05697870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05080211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05628658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519923 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05500489 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05482347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05449116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519907 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05688191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05382854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05467243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24519974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05006240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24520003 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10538853 - -
          Affymetrix Mouse Gene 1.0 ST Array 10538854 - -
          Affymetrix HT MG-430 PM Array Plate 1440661_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10538852 - -
          Mouse Genome 430 2.0 Array 1440661_at - -
          Mouse Genome 430 2.0 Array 1440661_PM_at - -
          Mouse Expression Set 430 1440661_at - -
          Mouse Expression Set 430 1440661_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440661_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440661_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 530119 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104480463 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline