Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00621924_m1
          See other TMEM198 GE Assays ›
          Gene Symbol:
          Tmem198
          Gene Name:
          transmembrane protein 198
          Gene Aliases:
          A230078I05Rik, Tmem198-1, Tmem198a
          Chromosome Location:
          Chr.1: 75479532 - 75485693 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00621924_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem198
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177056.4 NP_796030.1 3 - 4 1128 65
          GenBank mRNA AK038957.1 - 3 - 4 1089 65
          AK082334.1 - 2 - 3 1388 65
          AK158863.1 - 3 - 4 1602 65
          BC059868.1 - 3 - 4 1093 6809137 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100000661.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem198

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          Wnt signaling pathway
          positive regulation of canonical Wnt signaling pathway
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354397 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354367 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354400 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354399 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354370 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354411 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354408 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354369 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354344 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354410 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354412 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354401 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354368 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354345 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967837 BC059868.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967837 BC046546.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967837 AK082334.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967837 BC042776.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967837 NM_177056.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1967835 AK082334.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 67321 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582569 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 483294 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 543049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41587 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146630 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967837 BC059868.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967837 BC046546.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967837 AK082334.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967837 BC042776.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967837 NM_177056.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1967835 AK082334.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354359 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354357 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354363 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354365 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354354 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354371 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354368 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354367 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354364 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354376 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354356 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354379 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20354366 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00106032 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146630 - -
          Whole Mouse Genome G4122A A_52_P423709 BC059868.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423709 AK082334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423709 AK158863.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423709 NM_177056.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P423709 AK038957.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P277128 BC059868.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P277128 BC046546.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P277128 AK082334.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P277128 BC042776.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P277128 NM_177056.4 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110775 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111654 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146630 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00025600 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00025584 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00025595 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00025583 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354345 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354344 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354370 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354412 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354369 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354410 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354411 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354408 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354368 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354397 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354401 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354399 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354400 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20354367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20354466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354353 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354385 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354379 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354375 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00365244 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354343 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01085677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354348 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00365243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354373 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354365 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00725060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354366 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00725034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00365225 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354363 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354360 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00025578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20354356 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10347667 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347666 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347663 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347665 - -
          Affymetrix Mouse Gene 1.0 ST Array 10347668 - -
          Affymetrix HT MG-430 PM Array Plate 1455192_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10347662 - -
          Mouse Genome 430 2.0 Array 1455192_PM_at - -
          Mouse Genome 430 2.0 Array 1455192_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319998_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 319998_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100005681.mm.1 NM_177056.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100005681.mm.1 NM_177056.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100005681.mm.1 TC0100000661.mm.1 MTA TC
          Mouse Expression Set 430 1455192_PM_at - -
          Mouse Expression Set 430 1455192_at - -
          Affymetrix HT Mouse Genome 430B Array 1455192_at - -
          Affymetrix HT Mouse Genome 430B Array 1455192_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 930882 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3162551 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3162551 - -
          mouse-6 v1.1 (Illumina) GI_31342696-I - -
          MouseRef-8 v2 Expression BeadChip ILMN_3162551 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3162551 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) GI_31342696-I - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline