Search
Search

Fstl4| Gene Symbol: Fstl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177059.3 | NP_796033.2 | 8 - 9 | 1200 | 88 | |
| GenBank mRNA | AF374459.1 | - | 8 - 9 | 1200 | 88 | |
| AK046350.1 | - | 8 - 9 | 1251 | 88 | ||
| AK081114.1 | - | 8 - 9 | 88 | |||
| AK220367.1 | - | 4 - 5 | 580 | 88 | ||
| BC132353.1 | - | 8 - 9 | 1177 | 40130630 | 88 | |
Set Membership: |
|