Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00622009_m1
          See other FBXW21 GE Assays ›
          Gene Symbol:
          Fbxw21
          Gene Name:
          F-box and WD-40 domain protein 21
          Gene Aliases:
          E330009P21Rik
          Chromosome Location:
          Chr.9: 109136887 - 109162022 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00622009_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxw21
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177069.3 NP_796043.1 9 - 10 1330 100
          GenBank mRNA AK087710.1 - 9 - 10 1331 100
          BC099397.1 - 9 - 10 1312 30936381 100

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003077.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxw21

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P457529 BC099397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P457529 NM_177069.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P457529 AK087710.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P457529 BC099397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P457529 NM_177069.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P457529 AK087710.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P06111 BC099397.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P06111 NM_177069.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_65_P06111 AK087710.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 209687 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874687 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 910314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 422029 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 237592 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 471404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628486 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59931 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 164410 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 11068 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100365 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115148 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P457529 BC099397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P457529 NM_177069.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P457529 AK087710.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P457529 BC099397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P457529 NM_177069.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P457529 AK087710.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P06111 BC099397.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P06111 NM_177069.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_65_P06111 AK087710.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26818681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26818660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26818674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26818679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26818662 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115148 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06922373 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100365 - -
          Whole Mouse Genome G4122A A_52_P457529 BC099397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P457529 NM_177069.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P457529 AK087710.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P457529 BC099397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P457529 NM_177069.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P457529 AK087710.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P422894 BC099397.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P422894 AK136136.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P422894 NM_177069.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P422894 AK087710.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P422894 AK087902.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P109102 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115148 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100365 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07671347 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07582757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818522 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26818627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07943353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818677 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08147087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818690 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818632 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818641 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07596263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07827257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26818685 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10596993 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596999 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596998 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596994 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596991 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596997 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596992 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596996 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596995 - -
          Affymetrix HT MG-430 PM Array Plate 1441569_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10596990 - -
          Mouse Genome 430 2.0 Array 1441569_PM_at - -
          Mouse Genome 430 2.0 Array 1441569_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320082_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 320082_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900025932.mm.1 NM_177069.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025932.mm.1 NM_177069.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025932.mm.1 TC0900003077.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025934.mm.1 NM_177069.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025934.mm.1 NM_177069.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025934.mm.1 TC0900003077.mm.1 MTA TC
          Mouse Expression Set 430 1441569_PM_at - -
          Mouse Expression Set 430 1441569_at - -
          Murine Genome U74v2 Set 130550_PM_f_at - -
          Murine Genome U74v2 Set 130550_f_at - -
          Affymetrix HT Mouse Genome 430B Array 1441569_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1441569_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 594528 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1660341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1252646 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252646 - -
          mouse-6 v1.1 (Illumina) scl35340.8.1_0-S - -
          Illumina Sentrix mouse-6.v1 bead array 1660341 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35340.8.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1252646 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252646 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline