Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00622042_m1
          See other ADRBK2 GE Assays ›
          Gene Symbol:
          Adrbk2
          Gene Name:
          adrenergic receptor kinase, beta 2
          Gene Aliases:
          4833444A01Rik, AI851927, AW551196, Adrbk-2, Bark-2, GRK3
          Chromosome Location:
          Chr.5: 112910478 - 113015951 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00622042_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Adrbk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001285806.1 NP_001272735.1 4 - 5 465 62
          NM_177078.4 NP_796052.2 4 - 5 426 62
          GenBank mRNA AK048763.1 - 4 - 5 391 62
          AK076533.1 - 4 - 5 324 62
          AK134672.1 - 4 - 5 403 62
          AK141186.1 - 4 - 5 444 62
          AK172513.1 - 4 - 5 378 62
          CJ104587.1 - 4 - 5 427 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003024.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Adrbk2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          desensitization of G-protein coupled receptor protein signaling pathway
          protein phosphorylation
          intracellular protein transport
          signal transduction
          phosphorylation
          receptor internalization
          termination of G-protein coupled receptor signaling pathway
          inositol phosphate metabolic process
          rhodopsin metabolic process
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          G-protein coupled receptor kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          D1 dopamine receptor binding
          beta-adrenergic receptor kinase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500026888.mm.1 TC0500003024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026888.mm.1 NM_001285806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026890.mm.1 TC0500003024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026890.mm.1 NM_001285806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026890.mm.1 NM_177078.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026891.mm.1 TC0500003024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026891.mm.1 NM_001285806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026891.mm.1 NM_177078.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026892.mm.1 TC0500003024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026892.mm.1 NM_001285806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026892.mm.1 NM_177078.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026887.mm.1 TC0500003024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026887.mm.1 NM_001285806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026887.mm.1 NM_177078.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026889.mm.1 TC0500003024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026889.mm.1 NM_001285806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026889.mm.1 NM_177078.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026886.mm.1 TC0500003024.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500026886.mm.1 NM_001285806.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500026886.mm.1 NM_177078.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 494986 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline